Skip to Content
Merck
All Photos(1)

Key Documents

Q3625

Sigma-Aldrich

Quinidine

sodium channel blocker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2
CAS Number:
Molecular Weight:
324.42
Beilstein:
91866
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Quinidine, anhydrous

grade

anhydrous

impurities

≤20% Dihydroquinidine (according to USP specifications., actual content given on label)

mp

168-172 °C (lit.)

originator

Bayer

SMILES string

COc1ccc2nccc([C@H](O)C3CC4CCN3C[C@@H]4C=C)c2c1

InChI

1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19+,20-/m0/s1

InChI key

LOUPRKONTZGTKE-LHHVKLHASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Quinidine has been used:
  • as an inhibitor of cytochrome P450 enzyme isoform CYP2D6[1]
  • as a potassium (K+) channel blocker in microglia[2]
  • to test its effect on electrophysiological behaviour of human induced pluripotent stem cells (hiPSC) cardiomyocytes[3]

Biochem/physiol Actions

Class IA antiarrhythmic; potassium channel blocker.
Quinidine, an isomer of quinine has potential side effects like proarrhythmia on usage. It prolongs cardiac potential and is sodium channel blocker.[4] It is a potential antagonist for α1-adrenoceptors and contributes to hypotension.[5] It inhibits cytochrome P450 2D6 and improves circulation and brain penetration of dementia drug, dextromethorphan.[6] Quinidine-dextromethorphan combination may be useful in treating pseudobulbar affect (PBA).[7]

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of quinidine and verapamil on human cardiovascular alpha1-adrenoceptors
Shibata K, et al.
Circulation, 97(13), 1227-1230 (1998)
Microglial ramification, surveillance, and interleukin-1beta release are regulated by the two-pore domain K+ channel THIK-1
Madry C, et al.
Neuron, 97(2), 299-312 (2018)
Dextromethorphan/quinidine: a review of its use in adults with pseudobulbar affect
Yang LPH and Deeks ED
Drugs, 75(1), 83-90 (2015)
Cardiac Sodium Channel Disorders An Issue of Cardiac Electrophysiology Clinics E-Book, 6(4) (2015)
Pharmacological evaluation of C-3 modified Betulinic acid derivatives with potent anticancer activity
Rajendran P, et al.
Investigational New Drugs, 26(1), 25-34 (2008)

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Protocols

LC/MS/MS Analysis of Interacting Cardiac Drugs Digoxin, Quinidine, Amiodarone and Verapamil on Titan™ C18

LC/MS/MS Analysis of Interacting Cardiac Drugs Digoxin, Quinidine, Amiodarone and Verapamil on Titan™ C18

LC/MS/MS Analysis of Interacting Cardiac Drugs Digoxin, Quinidine, Amiodarone and Verapamil on Titan™ C18

LC/MS/MS Analysis of Interacting Cardiac Drugs Digoxin, Quinidine, Amiodarone and Verapamil on Titan™ C18

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service