Skip to Content
Merck
All Photos(3)

Key Documents

M107

Sigma-Aldrich

(+)-MK-801 hydrogen maleate

powder

Synonym(s):

(5S,10R)-(+)-5-Methyl-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5,10-imine hydrogen maleate, Dizocilpine hydrogen maleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N · C4H4O4
CAS Number:
Molecular Weight:
337.37
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

form

powder

Quality Level

optical activity

[α]25/D +117°, c = 1 in methanol(lit.)

color

white

solubility

DMSO: >20 mg/mL
H2O: 6 mg/mL
0.1 M HCl: soluble
ethanol: soluble

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.C[C@]12N[C@H](Cc3ccccc13)c4ccccc24

InChI

1S/C16H15N.C4H4O4/c1-16-13-8-4-2-6-11(13)10-15(17-16)12-7-3-5-9-14(12)16;5-3(6)1-2-4(7)8/h2-9,15,17H,10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t15-,16+;/m1./s1

InChI key

QLTXKCWMEZIHBJ-PJGJYSAQSA-N

General description

(+)-MK-801 hydrogen maleate is a highly potent and selective non-competitive NMDA glutamate receptor antagonist that acts at the NMDA receptor-operated ion channel as an open channel blocker. Neurons are depolarized when MK-801 binds to the Glu (40 Glu) receptor (NMDA receptor- ion channel complex) at two sites. 40 Glu is the brain′s primary excitatory neurotransmitter. MK-801 attaches to the ion channel at the binding site PCP of the receptor. Receptor blockade occurs in a voltage-dependent manner. It also known as dizocilpine hydrogen maleate is an effective anticonvulsant with both anxiolytic and sympathomimetic properties.(+)-MK-801 hydrogen maleate stimulates locomotion in monoamine-depleted mice.

Application

(+)-MK-801 hydrogen maleate has been used in following experimental studies:
It is injected to mice to study the effect of systemic administration of CIQ [(3-chlorophenyl) (6, 7-dimethoxy-1-((4-methoxyphenoxy) methyl)-3, 4-dihydroisoquinolin-2(1H)-yl)methanone] on schizophrenia-like behaviours stimulated by MK-801.
It has been used to block the NMDA (N-methyl-D-aspartate) receptor-mediated rtPA (recombinant tissue plasminogen activator)-enhanced excitotoxic pathway.
It is injected in varied doses to study behavioral performance altering effects of MK-801 in zebrafish (Danio rerio).
It is used as a NMDA receptor antagonist to study 3-OH-GA-induced neuronal damage via NMDA receptors.

Biochem/physiol Actions

Experimental study shows that MK-801, influence circling behavior in a direction-specific manner, modify swimming activity and place preference in zebrafish. Acute injection of MK-801, dose dependently, enhances extracellular acetylcholine (ACh) release in the rat parietal cortex. For intracerebral hematomas, MK-801 might act as a potent supplement to reduce delayed edema after recombinant tissue plasminogen activator (rtPA) clot lysis. MK-801 functions as an antidepressant in the social defeat stress model, but it doesn’t have long-lasting effect.
Highly potent and selective non-competitive NMDA glutamate receptor antagonist that acts at the NMDA receptor-operated ion channel as an open channel blocker. Inhibits behavioral sensitization to psychostimulants and ethanol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth K Ruzzo et al.
Cell, 178(4), 850-866 (2019-08-10)
We performed a comprehensive assessment of rare inherited variation in autism spectrum disorder (ASD) by analyzing whole-genome sequences of 2,308 individuals from families with multiple affected children. We implicate 69 genes in ASD risk, including 24 passing genome-wide Bonferroni correction and
Management of delayed edema formation after fibrinolytic therapy for intracerebral hematomas: preliminary experimental data.
Rohde V
Acta Neurochirurgica. Supplement, 105, 101-104 (2008)
Gabrielle Girardeau et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(15), 5176-5183 (2014-04-11)
Hippocampal sharp wave-ripples (SPW-Rs) and associated place-cell reactivations are crucial for spatial memory consolidation during sleep and rest. However, it remains unclear how learning and consolidation requirements influence and regulate subsequent SPW-R activity. Indeed, SPW-R activity has been observed not
Behavioral performance altering effects of MK-801 in zebrafish (Danio rerio).
Sison M and Gerlai R.
Behavioural Brain Research, 220(2), 331-337 (2011)
The NMDA antagonist MK-801 causes marked locomotor stimulation in monoamine-depleted mice.
Carlsson M and Carlsson A
Journal of Neural Transmission. General Section, 75(3), 221-226 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service