Skip to Content
Merck
All Photos(1)

Key Documents

N6658

Sigma-Aldrich

Nutrient Mixture F-12 Ham

With ʟ-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Ham’s F-12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

Nutrient Mixture F-12 Ham, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

L-glutamine: yes
HEPES: no
phenol red: yes
sodium pyruvate: 0.11 g/L
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Ham′s Nutrient Mixtures were mainly created to support the clonal growth of numerous clones of Chinese hamster ovary (CHO) cells and clones of HeLa and mouse L-cells. It is also used to grow primary rat hepatocytes and rat prostate epithelial cells. Ham′s F-12 is used as the medium of choice for a clonal toxicity assay using CHO cells.

Application

Nutrient Mixture F-12 Ham has been used:
  • to culture Chinese hamster ovary cells overexpressing insulin receptor (CHO-IR)
  • to culture LAN-1 cells
  • to maintain SUM149, SUM159, SUM190, FC-IBC02, BCX010, and IBC3 cells
  • with Dulbecco’s modified Eagle’s medium (DMEM) media in a 1:1 ratio to culture oral cavity squamous cell carcinoma (OSCC) cell lines.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianjia Fan et al.
Journal of lipid research, 59(5), 830-842 (2018-03-23)
apoE is the primary lipid carrier within the CNS and the strongest genetic risk factor for late onset Alzheimer's disease (AD). apoE is primarily lipidated via ABCA1, and both are under transcriptional regulation by the nuclear liver X receptor (LXR).
Ali I Mohammed et al.
Biomolecules, 13(8) (2023-08-26)
Reactive oxygen species (ROS) are highly reactive molecules generated in living organisms and an excessive production of ROS culminates in oxidative stress and cellular damage. Notably, oxidative stress plays a critical role in the pathogenesis of a number of oral
Małgorzata Durbas et al.
Apoptosis : an international journal on programmed cell death, 23(9-10), 492-511 (2018-07-22)
The process of autophagy and its role in survival of human neuroblastoma cell cultures was studied upon addition of an anti-GD2 ganglioside (GD2) 14G2a mouse monoclonal antibody (14G2a mAb) and an aurora A kinase specific inhibitor, MK-5108. It was recently
Heidi Olzscha et al.
Cell chemical biology, 24(1), 9-23 (2016-12-19)
Lysine acetylation is becoming increasingly recognized as a general biological principle in cellular homeostasis, and is subject to abnormal control in different human pathologies. Here, we describe a global effect on amyloid-like protein aggregation in human cells that results from
Michelle J Nyhan et al.
BMC cancer, 16, 101-101 (2016-02-18)
Successful treatment of oesophageal cancer is hampered by recurrent drug resistant disease. We have previously demonstrated the importance of apoptosis and autophagy for the recovery of oesophageal cancer cells following drug treatment. When apoptosis (with autophagy) is induced, these cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service