Skip to Content
Merck
All Photos(1)

Documents

M8642

Sigma-Aldrich

Anti-Mouse IgG (whole molecule) antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-mouse antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

200
300

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

Ouchterlony double diffusion: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections
Anti-Mouse IgG (whole molecule) antibody recognizes IgG1, G2a, G2b, and G3, mouse IgM and IgA. The identity and purity of affinity isolated antibody is established by immunoelectrophoresis.

Immunogen

Pooled normal mouse serum

Application

Anti-Mouse IgG (whole molecule) antibody was used as secondary antibody in immunoblotting at a working dilution of 1:1000. The antibody was used in ELISA and RIA in various studies.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)

Physical form

Lyophilized from 0.01 M sodium phosphate, 0.015 M sodium chloride, pH 7.2

Reconstitution

Reconstitute with 0.135 M sodium chloride.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristian M Forbes et al.
PloS one, 9(3), e91113-e91113 (2014-03-14)
Marked variation occurs in both seasonal and multiannual population density peaks of northern European small mammal species, including voles. The availability of dietary proteins is a key factor limiting the population growth of herbivore species. The objective of this study
Govert J van Dam et al.
Experimental parasitology, 135(2), 274-282 (2013-07-16)
An earlier reported laboratory assay, performed in The Netherlands, to diagnose Schistosoma infections by detection of the parasite antigen CAA in serum was converted to a more user-friendly format with dry reagents. The improved assay requires less equipment and allows
Orhan Kankavi
Acta biochimica Polonica, 50(4), 1057-1064 (2004-01-24)
The presence of surfactant proteins was investigated in the human organ of Corti, Eustachian tube and kidney tissues. It has previously been shown that lamellar bodies are present in hairy cells of organ of Corti, in the cytoplasm of secretory
Jusung Oh et al.
Theranostics, 8(12), 3189-3197 (2018-06-23)
The immunochromatographic (ICA) assay is a highly promising platform for rapid and simple detection of C-reactive protein (CRP) which is an indicator of the different phases of various diseases, as well as of inflammation and infection. However, the hook effect
Samuel Roulin et al.
Planta, 215(1), 51-59 (2002-05-16)
When seedlings of barley (Hordeum vulgare L.) were transferred from a natural light/dark cycle into darkness, (1-->3,1-->4)-beta- D-glucan endohydrolase (EC 3.2.1.73) activity in leaf extracts increased 3- to 4-fold after 2 days. Activity decreased to normal levels within a day

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service