Skip to Content
Merck
All Photos(1)

Key Documents

M6898

Sigma-Aldrich

Anti-Mouse IgG (Fab specific) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Mouse Fab Antibody, Anti-Mouse Fab Antibody - Anti-Mouse IgG (Fab specific) antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect ELISA: 1:20,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Goat polyclonal anti-Mouse IgG (Fab specific) reacts with normal mouse serum, mouse IgG and the Fab fragment of mouse IgG, while no reaction is observed against the Fc fragment of mouse IgG. No reaction is observed with human IgG.
Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Mice possess five immunoglobulin classes- IgM, IgG, IgA, IgD and IgE. Mouse IgG is further divided into five classes- IgG1, IgG2a, IgG2b, IgG2c and IgG3.
Mouse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in mouse serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.

Application

Anti-Mouse IgG (Fab specific) antibody produced in goat has been used in western blotting.
Goat polyclonal anti-Mouse IgG (Fab specific) may be used for the detection and quantitation of mouse IgG in serum and biological fluids. It may be used to monitor changes in IgG levels under various conditions such as infection. It may also be used when conjugated with a detector molecule as a secondary antibody.

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III reactions. IgG2a has cytotropic properties and IgG2 molecules have the ability to activate the complement system. IgG helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Other Notes

Antibody adsorbed with human IgG.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan-Wei Sha et al.
Clinical genetics, 95(2), 277-286 (2018-10-10)
The majority of men with defects in spermatogenesis remain undiagnosed. Acephalic spermatozoa is one of the diseases causing primary infertility. However, the causes underlying over half of affected cases remain unclear. Here, we report by whole-exome sequencing the identification of
Deficiency of gap junction composed of connexin43 contributes to oxaliplatin resistance in colon cancer cells
Su M and Zhang Q
Oncology Letters, 14(3), 3669-3674 (2017)
Connexin 43 expressed in endothelial cells modulates monocyte-endothelial adhesion by regulating cell adhesion proteins
Yuan D, et al.
Molecular Medicine Reports, 12(5), 7146-7152 (2015)
Keisuke Tonouchi et al.
International immunology, 32(9), 613-621 (2020-06-07)
Influenza A subtypes are categorized into group 1 and group 2 based on the hemagglutinin (HA) sequence. Owing to the phylogenetic distance of HAs in different groups, antibodies that bind multiple HA subtypes across different groups are extremely rare. In
Bahar Dasgeb et al.
Molecular imaging, 13 (2014-07-16)
Basal cell carcinoma (BCC), the most common cancer in humans, appears macroscopically and microscopically similar to many other skin lesions, which makes differential diagnosis difficult. We are developing an approach for quantitative molecular imaging of BerEP4, a transmembrane biomarker for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service