Skip to Content
Merck
All Photos(2)

Key Documents

A0168

Sigma-Aldrich

Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat

affinity isolated antibody

Synonym(s):

Goat Anti-Mouse IgG (Fc specific)–HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

direct ELISA: 1:50,000
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100
western blot: 1:80,000-1:160,000 using total cell extract of HeLa cells (5-10 μg per well

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases. Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice.
Antibody is isolated from goat anti-mouse IgG antiserum by immunospecific purification to remove essentially all goat serum proteins, including immunoglobulins that do not specifically bind to the Fc fragment of mouse IgG. Anti-Mouse IgG is conjugated to peroxidase by protein cross linking with 0.2% glutaraldehyde. The antibody preparation is solid phase adsorbed with human IgG to ensure minimal cross reactivity in tissue or cell preparations.

Immunogen

purified mouse IgG Fc fragment

Application

Anti-Mouse IgG (Fc specific)-Peroxidase antibody produced in goat has been used in
  • western blot
  • enzyme linked immunosorbent assay (ELISA) and
  • indirect immunoperoxidase assay (IIPA)

Specificity of a new antibody for PND in blood from immunized mice was tested by Elisa using biotynlated PND and streptavidin coated plates. HRP conjugated goat anti-mouse IgG (Fc specific) was used as secondary.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT.

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human antibodies targeting CD30+ lymphomas
Wezler X, et al.
Human Antibodies, 21(1-2), 13-28 (2012)
Intrathecal synthesis of specific antibodies in patients with invasion of the central nervous system by Mycoplasma pneumoniae.
Benvcina D, et al.
European Journal of Clinical Microbiology & Infectious Diseases : Official Publication of the European Society of Clinical Microbiology, 19(7), 521-530 (2000)
Structural insights of proteins in sub-cellular compartments: In-mitochondria NMR
Barbieri L, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1843(11), 2492-2496 (2014)
Identification of peptidases in Nicotiana tabacum leaf intercellular fluid
Delannoy M, et al.
Proteomics, 8(11), 2285-2298 (2008)
Anti-complement component 5 antibody targeting MG4 domain inhibits choroidal neovascularization
Jo DH, et al.
Testing, 8(28), 45506-45506 (2017)

Articles

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service