Skip to Content
Merck
All Photos(1)

Key Documents

R2004

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule) antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Whole antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

immunoelectrophoresis: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Binds all rabbit Igs.

Specificity

Anti-Rabbit IgG (whole molecule) antibody detects all rabbit Igs.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Physical form

Lyophilized from 0.01 M sodium phosphate, 0.015 M sodium chloride, pH 7.2

Reconstitution

Reconstitute with 0.135 M sodium chloride.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brinky Desai et al.
Conservation physiology, 12(1), coae035-coae035 (2024-06-06)
Animals face several challenges in their natural environment, and to cope with such conditions, they may exhibit contrasting physiological responses that directly affect their overall well-being and survival. In this study, we assessed physiological responses via faecal glucocorticoid metabolite (fGCM)
Nina Kirstein et al.
eLife, 10 (2021-03-09)
Eukaryotic DNA replication initiates during S phase from origins that have been licensed in the preceding G1 phase. Here, we compare ChIP-seq profiles of the licensing factors Orc2, Orc3, Mcm3, and Mcm7 with gene expression, replication timing, and fork directionality
Xuefeng Wang et al.
Biosensors & bioelectronics, 26(5), 1871-1875 (2010-03-20)
Fluorescence-free biosensor arrays for protein detection directly measure the protein surface density, and do not require a fluorophore or enzyme label, and provide quantitative and consistent signals. However, few fluorescence-free biosensor protein arrays have demonstrated successful application in high-background samples
Sung Eun Seo et al.
Sensors and actuators. B, Chemical, 381, 133364-133364 (2023-01-24)
Since December 2019, the rapid and sensitive detection of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has become a priority for public health. Although the lateral flow assay (LFA) sensor has emerged as a rapid and on-site SARS-CoV-2 detection technique
Ishwar Gill et al.
Journal of cell science, 128(4), 768-780 (2014-12-21)
During cortical development, N-methyl-D-aspartate (NMDA) receptors (NMDARs) facilitate presynaptic terminal formation, enhance neurotransmitter release and are required in presynaptic neurons for spike-timing-dependent long-term depression (tLTD). However, the extent to which NMDARs are found within cortical presynaptic terminals has remained controversial

Articles

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service