Skip to Content
Merck
All Photos(1)

Documents

A9316

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Goat Anti-Mouse IgG (whole molecule)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

technique(s)

direct ELISA: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Goat polyclonal anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody is reactive with mouse IgG1, IgG2a, IgG2b, IgG3, IgA, and IgM. Cross reactivity of the antibody conjugate is determined by ELISA. The conjugate shows no reactivity with rat IgG or rat serum proteins.
Immunoglobulins (Igs) belong to the immunoglobulin super-family. IgG is an abundant immunoglobulin present in human serum. The four classes of IgG are IgG1, IgG2, IgG3, and IgG4.
Mouse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in mouse serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.

Application

Anti-Mouse IgG (whole molecule) Alkaline Phosphatase antibody produced in goat has been used as a secondary antibody in:
  • histological studies in heart and liver grafts
  • immunohistochemical studies of gangliosides and immunostaining of lymphoid organs
  • western blot detection in blood samples
Surfactant Protein A was detected in bronchoalveolar fluid using alkaline phosphatase conjugated goat anti-mouse IgG as the secondary at μg/ml in TBS/Tween containing final concentration of 0.5M NaCl.
The level of antibodies specific to P. aeuginosa present in immunized mouse sera was determined by ELISA using alkaline phosphatase-conjugated goat anti-mouse IgG as the secondary antibody at a dilution of 1:1000.

Biochem/physiol Actions

IgG1 deficiency results in hypogammaglobulinemia. IgG2 deficiency increases susceptibility to bacterial infections. IgG3 mediates effector functions and IgG4 is associated with asymptomatic infection.

Other Notes

Antibody adsorbed with rat serum proteins.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 10 mM glycine, 1% bovine serum albumin, 1 mM MgCl2, 50% glycerol and 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background with rat samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Significant enhancement by anti-ICOS antibody of suboptimal tacrolimus immunosuppression in rat liver transplantation
Guo L, et al.
Liver Transplantation, 10(6), 743-747 (2004)
Prolonged Cardiac Allograft Survival In Rats Systemically Injected Adenoviral Vectors Containing Ctla4ig-gene1
Kita Y, et al.
Transplantation, 68(6), 758-766 (1999)
Alcohol administration blocks stress-induced impairments in memory and anxiety, and alters hippocampal neurotransmitter receptor expression in male rats
Gomez JL, et al.
Hormones and Behavior, 63(4), 659-666 (2013)
Elizabeth A García et al.
Veterinary microbiology, 222, 30-38 (2018-08-07)
Mycobacterium bovis is the causative agent of bovine tuberculosis and is a member of Mycobacterium tuberculosis complex, which causes tuberculosis in a number of mammals including humans. Previous studies have shown that the genes encoding the two-component system PhoPR, which
The microstructure of secondary lymphoid organs that support immune cell trafficking
Matsuno K, et al.
Archives of Histology and Cytology, 73(1), 1-21 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service