Skip to Content
Merck
All Photos(1)

Key Documents

M6418

Sigma-Aldrich

Manumycin A from Streptomyces parvulus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H38N2O7
CAS Number:
Molecular Weight:
550.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥97% (HPLC)

Quality Level

form

powder

solubility

methanol: 9.80-10.20 mg/mL, clear, yellow to very deep yellow

storage temp.

2-8°C

SMILES string

CCCC[C@@H](C)\C=C(C)\C=C(/C)C(=O)NC1=C[C@@](O)(\C=C\C=C\C=C\C(=O)NC2=C(O)CCC2=O)[C@@H]3O[C@H]3C1=O

InChI

1S/C31H38N2O7/c1-5-6-11-19(2)16-20(3)17-21(4)30(38)32-22-18-31(39,29-28(40-29)27(22)37)15-10-8-7-9-12-25(36)33-26-23(34)13-14-24(26)35/h7-10,12,15-19,28-29,34,39H,5-6,11,13-14H2,1-4H3,(H,32,38)(H,33,36)/b8-7+,12-9+,15-10+,20-16+,21-17+/t19-,28?,29-,31+/m1/s1

InChI key

TWWQHCKLTXDWBD-YCWSEAROSA-N

Application

Manumycin A from Streptomyces parvulus has been used to inhibit IκB kinase (IKK)b−nuclear κ-B essential modulator (NEMO) interaction in the homogeneous time-resolved fluorescence (HTRF)-based binding assay.

Biochem/physiol Actions

Manumycin A is a natural monomeric epoxyquinoid. It has an ability to inhibit tumor necrosis factor (TNF) induced IκB kinase (IKK) activity in various cell types. In addition, manumycin A exhibits anti-tumor property by inhibiting farnesylation of oncogenic Ras.
Potent inhibitor of Ras farnesyltransferase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hara et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(6), 2281-2285 (1993-03-15)
A microbial screen using a yeast strain with conditional deficiency in the GPA1 gene was carried out to search for inhibitors of protein farnesyltransferase (PFT). A strain of Streptomyces was found to produce active compounds named UCF1-A, UCF1-B, and UCF1-C.
Binding of manumycin A inhibits IkappaB kinase beta activity
Bernier M, et al.
Test, 281(5), 2551-2561 (2006)
William Stein et al.
The Journal of heart and lung transplantation : the official publication of the International Society for Heart Transplantation, 30(7), 761-769 (2011-04-05)
In this study we systematically dissect the prenylation pathway to better define the mechanism behind statin inhibition in chronic allograft rejection in heart transplants, or transplant coronary artery disease (TCAD). Utilizing a murine heterotopic heart transplant model, animals received daily
Qingming Xiong et al.
Journal of medical microbiology, 60(Pt 6), 744-749 (2011-02-26)
Anaplasma phagocytophilum is an obligately intracellular bacterium and is the causative agent of human granulocytic anaplasmosis (HGA), an emerging and major tick-borne disease in the USA and other parts of the world. This study showed that the prenylation inhibitor manumycin
P K Singha et al.
Cell death & disease, 4, e457-e457 (2013-01-19)
Therapy resistance can be attributed to acquisition of anti-apoptotic mechanisms by the cancer cells. Therefore, developing approaches that trigger non-apoptotic cell death in cancer cells to compensate for apoptosis resistance will help to treat cancer effectively. Triple-negative breast cancers (TNBC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service