Skip to Content
Merck
All Photos(1)

Key Documents

P8340

Sigma-Aldrich

Protease Inhibitor Cocktail

Protease Inhibitor Cocktail
1 of 4 reviewers received a sample product or took part in a promotion

DMSO solution, for the inhibition of serine, cysteine, acid proteases and aminopeptidases, for use with mammalian cell and tissue extracts, DMSO solution

Synonym(s):

Protease Inhibitor Mix

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
form:
DMSO solution
solubility:
water: soluble
storage temp.:
−20°C
Pricing and availability is not currently available.

Product Name

Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution

biological source

bovine lung

Quality Level

form

DMSO solution

solubility

water: soluble

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Protease Inhibitor Cocktail for mammalian cell and tissue extracts, in DMSO solution. It contains six individual components for inhibiting serine, cysteine, and acid proteases, and aminopeptidases.

Specificity

Inhibits serine, cysteine, and acid proteases, and aminopeptidases.

Application

Ideal for use as a buffer component during sonication, lysis buffer, and RIPA buffer

Provides protection against endogenous enzyme activity in cell and tissue extracts

Biochem/physiol Actions

This mixture contains individual components, including AEBSF at 104 mM, Aprotinin at 80 μM, Bestatin at 4 mM, E-64 at 1.4 mM, Leupeptin at 2 mM and Pepstatin A at 1.5 mM. Each component has specific inhibitory properties. AEBSF and Aprotinin act to inhibit serine proteases, including trypsin, chymotrypsin, and plasmin amongst others. Bestatin inhibits aminpeptidases. E-64 acts against cystein proteases. Leupeptin acts against both serine and cystein proteases. Pepstatin A inhibits acid proteases.

Features and Benefits

Broad-spectrum inhibition: inhibits multiple classes of proteases and aminopeptidases

Stable storage: supplied as a solution in DMSO, allowing for long-term storage at -20°C

Consistent performance: ready-to-use solution

Components

AEBSF
Aprotinin
Bestatin
E-64
Leupeptin
Pepstatin A

Caution

This cocktail should be stored at -20 °C.

Preparation Note

One mL of this cocktail solution is recommended for the inhibition of endogenous enzymes found in 100 mL of lysate from 20 g of bovine liver or 10 mL of lysate from CHO cells. This product is supplied as a solution in DMSO.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Signaling through extracellular signal-regulated kinase is required for spermatogonial proliferative response to stem cell factor.
Dolci S et al
The Journal of Biological Chemistry, 276(43), 40225-40233 (2001)
A transgenic rat that develops Alzheimer's disease-like amyloid pathology, deficits in synaptic plasticity and cognitive impairment.
Liu L et al
Neurobiology of Disease, 31(1), 46-57 (2008)
Interaction of Huntington disease protein with transcriptional activator Sp1.
Li SH et al
Molecular and Cellular Biology, 22(5), 1277-1287 (2002)
Megan D Schertzer et al.
Molecular cell, 75(3), 523-537 (2019-07-02)
Long noncoding RNAs (lncRNAs) cause Polycomb repressive complexes (PRCs) to spread over broad regions of the mammalian genome. We report that in mouse trophoblast stem cells, the Airn and Kcnq1ot1 lncRNAs induce PRC-dependent chromatin modifications over multi-megabase domains. Throughout the
Acceleration and persistence of neurofibrillary pathology in a mouse model of tauopathy following anesthesia.
Planel E et al
Faseb Journal, 23(8), 2595-2604 (2009)

Related Content

Research evaluating immune checkpoints may help identify those likely to benefit from PD-1/PD-L1 immunotherapy and reveal new immunotherapy targets for future investigation. See how the MILLIPLEX® Human Immuno-Oncology Checkpoint Protein Panel 2 was used to multiplex immune checkpoint molecules for NSCLC research.

Research evaluating immune checkpoints may help identify those likely to benefit from PD-1/PD-L1 immunotherapy and reveal new immunotherapy targets for future investigation. See how the MILLIPLEX® Human Immuno-Oncology Checkpoint Protein Panel 2 was used to multiplex immune checkpoint molecules for NSCLC research.

Research evaluating immune checkpoints may help identify those likely to benefit from PD-1/PD-L1 immunotherapy and reveal new immunotherapy targets for future investigation. See how the MILLIPLEX® Human Immuno-Oncology Checkpoint Protein Panel 2 was used to multiplex immune checkpoint molecules for NSCLC research.

Research evaluating immune checkpoints may help identify those likely to benefit from PD-1/PD-L1 immunotherapy and reveal new immunotherapy targets for future investigation. See how the MILLIPLEX® Human Immuno-Oncology Checkpoint Protein Panel 2 was used to multiplex immune checkpoint molecules for NSCLC research.

See All

Questions

1–10 of 15 Questions  
  1. Can this product be used for IL-6 detection using ELISA in rat tissue Homogenate? Can it be used along with PMSF without creating any problem in the assay?

    1 answer
    1. This product can be used in rat tissue homogenate. It already contains serine and cysteine protease inhibitors. Note that PMSF has a short half-life in aqueous solutions, specifically 55 minutes at 25°C and pH 7.5.

      Helpful?

  2. How long is the protease inhibitor cocktail stable at room temperature?

    1 answer
    1. This product ships without wet or dry ice and is stable for up to two weeks at ambient temperatures. The long-term stability of this material at room temperature has not been determined. The recommended long-term storage temperature is -20°C.

      Helpful?

  3. Can we dilute it in water? The experiment I plan to do will require 100ul of Protease inhibitor per vial, and I have more than 50 samples to run.

    1 answer
    1. This product is ready to use and should be diluted into the lysis buffer.

      Helpful?

  4. Is the protease cocktail P8340 supplied as a 100X solution or a 1X solution?

    1 answer
    1. This cocktail is essentially a 100X preparation. One milliliter of this solution is sufficient for the inhibition of endogenous enzymes found in 100 mL of lysate. Please see the link below to review this and additional information available in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/107/629/p8340dat-ms.pdf

      Helpful?

  5. What is the stock concentration of the Protease Inhibitor Cocktail P8340? Is it 1x or 100x?

    1 answer
    1. The concentration of inhibitors in this product is the following:AEBSF, 104 mMAprotinin, 0.085 mMBestatin, 4 mME-64, 1.4 mMLeupeptin, 2 mMPepstain A, 1.5 mM.

      Helpful?

  6. Do we need to add PMSF as well along with this cocktail? I intend to use this for RIPA buffer

    1 answer
    1. PMSF is a serine protease inhibitor. You can certainly add along with the cocktail if needed.

      Helpful?

  7. What is the working concentration if adding to the blood?

    1 answer
    1. The recommended amount to use is 1:100 (v:v) directly to the whole blood, but one has to take into consideration that this will disrupt the coagulation cascade. If it is a concern, the protease inhibitor cocktail should be added after separation of the blood components. In the serum use at the same concentration as for blood, 1:100 (v:v)

      There are articles citing this product when using blood samples. 1-100 was used in both articles:
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC7346264/
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC2722770/

      Helpful?

  8. How long can this product be stored at -20C?

    1 answer
    1. This product is not assigned an expiration date. It is assigned a recommended retest date of approximately 4 years after the quality release date. A recommended retest date is the period of time during which the product is expected to remain within established stability specifications, provided that it has been stored under defined conditions. The retest date can be found in the lot specific Certificate of Analysis. After the Retest Date, product samples should be examined to ensure that the product is still in compliance with the established specifications. For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/quality-services.

      Please see the link below to review a sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/P8340-1ML0000227406.pdf

      Helpful?

  9. Does Product P8340, Protease Inhibitor Cocktail, contain any chelators or EDTA?

    1 answer
    1. This product does not contain any metal chelators.

      Helpful?

  10. Is there anything in Product P8340, Protease Inhibitor Cocktail, that will cause downstream problems with RT-PCR?

    1 answer
    1. While the product should not cause any downstream problems with RT-PCR, we would advise to clean up the reaction with either DMN10 (GenElute™ Direct mRNA Miniprep Kit) or T9424 (TRI Reagent®) for  purification.

      Helpful?

1–10 of 15 Questions  

Reviews

1 of 4 reviewers received a sample product or took part in a promotion
1–2 of 4 Reviews  

Active Filters

  1. California
    • Review 1
    • Votes 2
    5 out of 5 stars.

    Sigma Protease Inhibitor Cocktail works well

    This Protease Inhibitor Cocktail is to protect your proteins from a wide range of proteases. It inhibites proteolytic activity that can degrade the protein you have spent days isolating. It is in a convenient water-soluble buffer that can be add directly to the lysis buffer.

    Helpful?

  2. india
    • Review 1
    • Votes 0
    5 out of 5 stars.

    It works very nicely for tissue lysate as well as cell lysate preperation.

    We always recommend using this product to obtain high quality western blot data.It is really nice.

    Helpful?

  3. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service