Skip to Content
Merck
All Photos(1)

Key Documents

C8999

Sigma-Aldrich

Calpeptin

≥98% (HPLC)

Synonym(s):

N-Benzyloxycarbonyl-L-leucylnorieucinal, Z-Leu-Nle-CHO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30N2O4
CAS Number:
Molecular Weight:
362.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)OCc1ccccc1)C=O

InChI

1S/C20H30N2O4/c1-4-5-11-17(13-23)21-19(24)18(12-15(2)3)22-20(25)26-14-16-9-7-6-8-10-16/h6-10,13,15,17-18H,4-5,11-12,14H2,1-3H3,(H,21,24)(H,22,25)/t17-,18-/m0/s1

InChI key

PGGUOGKHUUUWAF-ROUUACIJSA-N

General description

Calpeptin (Z-Leu-nLeu-H) is a cell permeable, peptide aldehyde inhibitor. It is more sensitive to calpain I compared to other inhibitors, such as Z-Leu-Met-H and leupeptin. Calpeptin stimulates neurite elongation in differentiating pheochromocytoma (PC12) cells. Calpeptin decreases microgliosis, astrogliosis, axonal damage and neuron and oligodendrocyte death in experimental autoimmune encephalomyelitis (EAE) spinal cord. Therefore, it can be considered as a potent therapeutic for treating EAE and multiple sclerosis (MS).

Application

Calpeptin has been used as a calpain inhibitor.

Biochem/physiol Actions

Calpeptin is a calpain inhibitor and rho kinase activator.
Calpeptin is a rho kinase activator and an inhibitor of calpains, a family of calcium-dependent cysteine proteases involved in apoptosis, long-term potentiation in neurons, and cell cycle progression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heike Hardelauf et al.
Lab on a chip, 11(16), 2763-2771 (2011-06-29)
Spatially defined neuronal networks have great potential to be used in a wide spectrum of neurobiology assays. We present an original technique for the precise and reproducible formation of neuronal networks. A PDMS membrane comprising through-holes aligned with interconnecting microchannels
Calpeptin, a calpain inhibitor, promotes neurite elongation in differentiating PC12 cells.
Pinter M, et al.
Neuroscience Letters, 170(1), 91-93 (1994)
Calpeptin attenuated inflammation, cell death, and axonal damage in animal model of multiple sclerosis.
Guyton M K, et al.
Journal of Neuroscience Research, 88(11), 2398-2408 (2010)
Hector N Aguilar et al.
PloS one, 6(6), e20903-e20903 (2011-06-23)
The 'phosphate-binding tag' (phos-tag) reagent enables separation of phospho-proteins during SDS-PAGE by impeding migration proportional to their phosphorylation stoichiometry. Western blotting can then be used to detect and quantify the bands corresponding to the phospho-states of a target protein. We
Inhibition of histone deacetylase 3 (HDAC3) mediates ischemic preconditioning and protects cortical neurons against ischemia in rats.
Yang X, et al.
Frontiers in Molecular Neuroscience, 9, 131-131 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service