Skip to Content
Merck
All Photos(1)

Key Documents

L5006

Sigma-Aldrich

Leucine Aminopeptidase, microsomal from porcine kidney

Type IV-S, ammonium sulfate suspension, 10-40 units/mg protein (Bradford)

Synonym(s):

Aminopeptidase M, Aminopeptidase N, Cathepsin III, LAP, Leucinamide aminopeptidase, Leucinaminopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type IV-S

Quality Level

form

ammonium sulfate suspension

specific activity

10-40 units/mg protein (Bradford)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Leucine Aminopeptidase belongs to the family of metalloprotease.

Application

Leucine Aminopeptidase, microsomal from porcine kidney has been used in the isolation of neprilysin (NEP) from rodent brain-C5 and for peptide digestion.

Biochem/physiol Actions

Leucine Aminopeptidase possesses broad substrate-specificity. It hydrolyzes peptides up to the proline residue and does not continue to degrade beyond proline.

Unit Definition

One unit will hydrolyze 1.0 μmole of L-leucine-p-nitroanilide to L-leucine and p-nitroaniline per min at pH 7.2 at 37 °C. (At 25 °C, approx. 40% of the activity at 37 °C is obtained.) The activity obtained using L-leucine p-nitroanilide as substrate is 2-5 times that obtained with L-leucinamide as substrate.

Physical form

Suspension in 3.5 M (NH4)2SO4 solution, pH 7.7, containing 10 mM MgCl2

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intestinal digestive resistance of immunodominant gliadin peptides
Hausch F, et al.
American Journal of Physiology: Gastrointestinal and Liver Physiology, 283(4), G996-G1003 (2002)
Biochemical identification of the neutral endopeptidase family member responsible for the catabolism of amyloid beta peptide in the brain
Takaki Y, et al.
The Journal of Biological Chemistry, 128(6), 897-902 (2000)
The most potent organophosphorus inhibitors of leucine aminopeptidase. Structure-based design, chemistry, and activity
Grembecka J, et al.
Journal of medicinal chemistry, 46(13), 2641-2655 (2003)
Rossella Galati et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 58(7-8), 558-561 (2003-08-27)
The ability of synthetic protein fragments to survive the degradative action of aminopeptidases and serum proteolytic enzymes can be remarkably enhanced by slight modifications at their N-terminal alpha-amino group. This can be achieved by addition of beta-alanine or amino acids
Debittering of a Tryptic Digest of Bovine p-casein Using Porcine Kidney General Aminopeptidase and X-Prolydipeptidyl Aminopeptidase from Lactococcus lactis subsp. cremoris AM2
Barry CM, et al.
Journal of Food Science, 65(7), 1145-1150 (2000)

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service