Skip to Content
Merck
All Photos(1)

Key Documents

A9934

Sigma-Aldrich

Aminopeptidase I from Streptomyces griseus

lyophilized powder, ≥200 units/mg protein

Synonym(s):

Leucine Aminopeptidase IV

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥200 units/mg protein

mol wt

21 kDa by gel filtration
33 kDa by SDS-PAGE

composition

Protein, 40-60% Lowry

storage temp.

−20°C

General description

Aminopeptidase I from Streptomyces griseus is a thermostable enzyme with Glu131 and Tyr246 as key active site residues.

Application

Aminopeptidase I from Streptomyces griseus has been used:
  • to test the biochar exposure effect on the enzyme activity
  • in circular dichroism (CD) spectroscopy studies
  • as a positive control in p-nitroanilide degradation assay

Biochem/physiol Actions

Aminopeptidase I from S. griseus has a fairly broad specificity, being able to remove the N-terminal residue of most proteins, except where the penultimate residue is an imino acid. It contains two Zn2+ binding sites. Aminopeptidase I from S. griseus is inhibited by 1,10-phenanthroline and is activated six-fold by Ca2+, which also stabilizes it against heat inactivation. This monomeric zinc metalloprotein has an isoelectric point (pI) of 5.4.
Aminopeptidase I may also be used as a reagent in the assay of endoprotease activities with a synthetic substrate in a two-stage assay. In the first stage, the endoprotease cleaves a peptide, such as Z-Y-X-Leu-p-nitroanilide, with the X, Y, and Z residues being chosen according to the specificity of the endoprotease.

Packaging

Package size based on protein content.

Unit Definition

One unit will hydrolyze 1.0 μmole of L-leucine-p-nitroanilide to L-leucine and p-nitroaniline per min at pH 8.0, 25 °C and 3.0 mM substrate concentration.

Physical form

Contains calcium acetate

Preparation Note

Reconstitute in 20 mM tricine, pH 8.0, with 0.05% bovine serum albumin. Dilute the enzyme with the reconstitution buffer to 0.15-0.3 U/mL for a working concentration. Solutions should be prepared fresh prior to use.

Other Notes

Endopeptidase contaminant: Not more than: 0.01 U/mg protein (as μmole tyrosine equivalent per min released from casein.)

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason L Parsons et al.
The FEBS journal, 272(22), 5753-5763 (2005-11-11)
Ionizing radiation, oxidative stress and endogenous DNA-damage processing can result in a variety of single-strand breaks with modified 5' and/or 3' ends. These are thought to be one of the most persistent forms of DNA damage and may threaten cell
Wakana Adachi et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 63(Pt 3), 200-203 (2007-03-03)
The vacuole hydrolase aminopeptidase 1 (Ape1) is a cargo protein transported to the vacuole by the cytosol-to-vacuole targeting (Cvt) pathway during conditions of growth and by autophagy during conditions of starvation. After transport to the vacuole, Ape1 is processed into
[Cytoplasm to vacuole targeting pathway in yeast].
Takahiro Shintani
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme, 51(10 Suppl), 1480-1483 (2006-08-23)
Bonnie K Baxter et al.
The Journal of biological chemistry, 280(47), 39067-39076 (2005-09-28)
The cytoplasm to vacuole (Cvt) trafficking pathway in S. cerevisiae is a constitutive biosynthetic pathway required for the transport of two vacuolar enzymes, aminopeptidase I (Ape1p) and alpha-mannosidase (Ams1p), to the vacuole. Ape1p and Ams1p bind to their receptor, Atg19p
Tomohiro Yorimitsu et al.
Molecular biology of the cell, 16(4), 1593-1605 (2005-01-22)
Proteins are selectively packaged into vesicles at specific sites and then delivered correctly to the various organelles where they function, which is critical to the proper physiology of each organelle. The precursor form of the vacuolar hydrolase aminopeptidase I is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service