Skip to Content
Merck
All Photos(1)

Key Documents

M6435

Sigma-Aldrich

Methionine Aminopeptidase from Pyrococcus furiosus

≥93% (SDS-PAGE), recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

Assay

≥93% (SDS-PAGE)

form

solution

specific activity

0.5 units/mg protein

mol wt

37 kDa by SDS-PAGE

UniProt accession no.

foreign activity

Other proteases, none detected

shipped in

dry ice

storage temp.

−20°C

Gene Information

Pyrococcus furiosus DSM 3638 ... PF0541(1468383)

Related Categories

General description

Methionine aminopeptidase from Pyrococcus furiosus is a 32 kDa thermostable enzyme.[1] It belongs to type 2a class of methionine aminopeptidase. Methionine aminopeptidase maintains protein homeostasis and coordinates posttranslational modification of proteins in eukaryotes.[2]
X-ray crystallography of the structure of methionine aminopeptidase from Pyrococcus furiosus or PfMAP was performed at a resolution of 1.75A and showed that the protein consists of a catalytic domain containing two cobalt ions in the active site and a unique insertion domain which is specific to the prokaryotic form of the protein. [3]

Application

Methionine Aminopeptidase from Pyrococcus furiosus has been used in a study to analyze the binding of Co(II)-specific inhibitors to the methionyl aminopeptidases from Escherichia coli and Pyrococcus furiosus. [4] It has also been used in a study to examine the binding of a new class of pseudopeptide analog inhibitors. [5]

Biochem/physiol Actions

Thermostable methionine aminopeptidase, which specifically liberates the N-terminal methioinine from proteins and peptides.

Unit Definition

One unit will hydrolyze 1 μmol of Met from Met-Pro-Ala-Ala-Gly in 1 minute at pH 7.5 at 37 °C.

Physical form

Solution containing 0.01% Tween® 20, 0.1 mM CoCl2, and 10 mM Tris-HCl, pH 7.5.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methionine aminopeptidase from the hyperthermophilic Archaeon Pyrococcus furiosus: molecular cloning and overexperssion in Escherichia coli of the gene, and characteristics of the enzyme
Tsunasawa S, et al.
Journal of Biochemistry, 122(4), 843-850 (1997)
Advances in bacterial methionine aminopeptidase inhibition
Helgren TR, et al.
Current Topics in Medicinal Chemistry, 16(4), 397-414 (2016)
A Ben-Bassat et al.
Journal of bacteriology, 169(2), 751-757 (1987-02-01)
Methionine aminopeptidase (MAP) catalyzes the removal of amino-terminal methionine from proteins. The Escherichia coli map gene encoding this enzyme was cloned; it consists of 264 codons and encodes a monomeric enzyme of 29,333 daltons. In vitro analyses with purified enzyme
A new colorimetric assay for methionyl aminopeptidases: Examination of the binding of a new class of pseudopeptide analog inhibitors
Mitra, S., et al.
Analytical Biochemistry, 357, 7-7 (2006)
Sanghamitra Mitra et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 14(4), 573-585 (2009-02-10)
Methionine aminopeptidases (MetAPs) represent a unique class of protease that is capable of the hydrolytic removal of an N-terminal methionine residue from nascent polypeptide chains. MetAPs are physiologically important enzymes; hence, there is considerable interest in developing inhibitors that can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service