Skip to Content
Merck
All Photos(3)

Key Documents

A8200

Sigma-Aldrich

Aminopeptidase from Aeromonas proteolytica

lyophilized powder, 50-150 units/mg protein

Synonym(s):

AAP, Aminopeptidase from Vibrio proteolyticus, bacterial leucyl aminopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

grade

Proteomics Grade

Quality Level

form

lyophilized powder

specific activity

50-150 units/mg protein

mol wt

29.5 kDa

composition

Protein, ~40% biuret

solubility

H2O: soluble 0.9-1.1 mg/mL, clear, colorless

foreign activity

endopeptidase, essentially free

storage temp.

−20°C

General description

A zinc-containing enzyme.

Specificity

Catalyzes the release of an N-terminal amino acid, preferentially leucine, but not glutamic or aspartic acids.

Application

Aminopeptidases are a family of widely distributed proteases, which may be used to study many significant biological processes such as protein maturation, hormone production, and peptide digestion. The enzyme has been used to measure the kinetic rate constant for the binding of bestatin, a general protease inhibitor, to aminopeptidase.[1]

Biochem/physiol Actions

Aminopeptidase from Aeromonas proteolytica is a metalloenzyme, which contains 2 atoms of Zn2+ in a single polypeptide with an approximate molecular weight of 29.5 kDa as determined by sedimentation. This enzyme has a high degree of stability, being stable even at temperatures of 70 °C for several hours. Partial inactivation occurs in 8 M urea. Maximum stability and activity are between pH 8.0-8.5. Aminopeptidase from Aeromonas proteolytica can function as an esterase.[2]
Aminopeptidase from Aeromonas proteolytica is involved in protein maturation, hormone production and peptide digestion.

Unit Definition

One unit will hydrolyze 1.0 μmole of L-leucine p-nitroanilide to L-leucine and p-nitroaniline per min at pH 8.0 at 25 °C.

Physical form

Lyophilized powder containing tricine buffer, pH 8.0, zinc chloride and stabilizer.

Preparation Note

Dissolves in water at 0.9-1.1 mg/mL concentration to form a clear, colorless solution.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James Kahn et al.
Biochemistry and molecular biology education : a bimonthly publication of the International Union of Biochemistry and Molecular Biology, 38(4), 238-241 (2011-05-14)
We have recently designed a biochemistry laboratory experiment for the purpose of providing students an advanced experience with enzyme kinetics and the kinetics of binding. Bestatin, a well-known and commercially available general protease inhibitor, is a slow-binding inhibitor of aminopeptidase
William T Desmarais et al.
Structure (London, England : 1993), 10(8), 1063-1072 (2002-08-15)
The aminopeptidase from Aeromonas proteolytica (AAP) is a bridged bimetallic enzyme that removes the N-terminal amino acid from a peptide chain. To fully understand the metal roles in the reaction pathway of AAP we have solved the 1.20 A resolution
C Schalk et al.
Archives of biochemistry and biophysics, 294(1), 91-97 (1992-04-01)
The heat-stable aminopeptidase from Aeromonas proteolytica has been purified using two new procedures, with the aim of preparing large single crystals for X-ray analysis. In a first procedure, we tried to avoid any drastic conditions capable of inducing microheterogeneities in
S Nirasawa et al.
The Biochemical journal, 341 ( Pt 1), 25-31 (1999-06-23)
An aminopeptidase from Aeromonas caviae T-64 was translated as a preproprotein consisting of three domains; a signal peptide (19 amino acid residues), an N-terminal propeptide (101 residues) and a mature region (273 residues). We demonstrated that a proteinase, which was
K M Huntington et al.
Biochemistry, 38(47), 15587-15596 (1999-11-26)
Peptide-derived thiols of the general structure N-mercaptoacyl-leucyl-p-nitroanilide (1a-c) were synthesized and found to be potent, slow-binding inhibitors of the aminopeptidase from Aeromonas proteolytica (AAP). The overall potencies (K(I)) of these inhibitors against AAP range from 2.5 to 57 nM exceeding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service