Skip to Content
Merck
All Photos(1)

Key Documents

S9761

Sigma-Aldrich

N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin

leucine aminopeptidase substrate, ≥98% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H39N5O9
CAS Number:
Molecular Weight:
661.70
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin, Leucine aminopeptidase substrate

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

methanol: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(NC(=O)CCC(O)=O)C(=O)NC(C)C(=O)N1CCCC1C(=O)NC(Cc2ccccc2)C(=O)Nc3ccc4C(C)=CC(=O)Oc4c3

InChI

1S/C34H39N5O9/c1-19-16-30(43)48-27-18-23(11-12-24(19)27)37-32(45)25(17-22-8-5-4-6-9-22)38-33(46)26-10-7-15-39(26)34(47)21(3)36-31(44)20(2)35-28(40)13-14-29(41)42/h4-6,8-9,11-12,16,18,20-21,25-26H,7,10,13-15,17H2,1-3H3,(H,35,40)(H,36,44)(H,37,45)(H,38,46)(H,41,42)

InChI key

FMVUZZZUYBXQGR-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Sohár et al.
Acta biologica Hungarica, 42(1-3), 265-274 (1991-01-01)
Previously, MMP-7ases were isolated from rat skeletal muscle by gel filtration and anion exchange chromatography. The enzyme that hydrolyzed succinyl-Ala-Ala-Pro-Phe-AMC (AMC: 7-amino-4-methyl-coumarin) was inhibited by EDTA. In this study we attempted to isolate MMP-7ase from mouse kidney. The isolation procedure
Julián Gamboa-Delgado et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 158(4), 251-258 (2010-12-21)
The effect of diet on larval growth, anionic trypsinogen gene expression (ssetryp1), and trypsin (EC 3.4.21.4) and chymotrypsin (EC 3.4.21.1) activities was assessed in Solea senegalensis. Changes in larval carbon stable isotope (δ(13)C) composition were used to estimate carbon assimilation.
Activity of cathepsin H,B and metalloproteinase in the serum of patients with acute myocardial infarction.
A László et al.
Clinica chimica acta; international journal of clinical chemistry, 210(3), 233-235 (1992-09-30)
G B Irvine et al.
Analytical biochemistry, 185(2), 304-307 (1990-03-01)
A simple, inexpensive, and sensitive assay for peptidase activity has been devised. The assay was performed in a microtiter plate and was based on fluorogenic peptide substrates, many of which are commercially available. 7-Amino-4-methyl coumarin the fluorescent product liberated during
H Sawada et al.
Experientia, 39(4), 377-378 (1983-04-15)
The action of a chymotrypsin-like enzyme from sperm extract from the ascidian Halocynthia roretzi was studied using several substrates. It was found that the most susceptible substrate had the most powerful inhibitory effect on fertilization in this animal. Among the

Articles

Analytical Enzyme Chymotrypsin: Chymotrypsin is produced in the acinar cells of the pancreas as the inactive precursor, chymotrypsinogen.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service