Skip to Content
Merck
All Photos(3)

Key Documents

L4418

Sigma-Aldrich

Anti-LSD1 (AOF2) (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Amine oxidase, flavin-containing, 2, Anti-BHC110, Anti-BRAF-HDAC complex protein 110, Anti-KIAA0601 protein, Anti-Lysine-specific demethylase 1, Anti-NPAO, Anti-Nuclear Polyamine Oxidase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 110-115 kDa

species reactivity

human, mouse (predicted), rat (predicted)

concentration

~1 mg/mL

technique(s)

immunocytochemistry: 5-10 μg/mL using MCF7 cells fixed with formaldehyde
immunoprecipitation (IP): 1-2 μg using MCF7 cell lysates
western blot: 1-2 μg/mL using MCF7 cell lysates (2 bands, 2 isoforms)

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AOF2(23028)
mouse ... Aof2(99982)

General description

Lysine specific demethylase 1 (LSD1) also known as amine oxidase flavin containing domain 2 (AOF2) is a lysine specific demethylase. It is a nuclear protein containing a Swi3p, Rsc8p and Moira (SWIRM) domain, a flavin adenine dinucleotide (FAD)-binding motif and an amine oxidase domain.

Immunogen

synthetic peptide corresponding to amino acids 835-852 of human LSD1 (AOF2) (isoform b), conjugated to KLH via an N-terminal added cysteine reisidue. The immunizing sequence is present also in isoform a. This sequence is conserved in human, rat, and mouse.

Application

Anti-LSD1 (AOF2) (C-terminal) antibody produced in rabbit immunoblotting, immunoprecipitation and immunofluorescence.

Biochem/physiol Actions

Lysine specific demethylase 1 (LSD1) is a component of several histone deacetylase complexes that function through modifying chromatin structure to repress transcription. LSD1 (AOF2) action can signal either activation or repression of transcription. Demethylation of histone H3 Lys4 by LSD1 (AOF2) leads to transcriptional repression of target genes, while demethylation of histone H3 at Lys9 leads to de-repression of androgen receptor target genes. The activity of LSD1 (AOF2) is regulated by its associated factors. REST corepressor 1 (CoREST), a transcriptional corepressor, promotes demethylation by enhancing the association of LSD1 (AOF2) with the nucleosomes, and it also protects LSD1 (AOF2) from proteasomal degradation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of LSD1 histone demethylase activity by its associated factors
Shi YJ, et al.
Molecular Cell, 19(6), 857-864 (2005)
LSD1 demethylates repressive histone marks to promote androgen-receptor-dependent transcription
Metzger E, et al.
Nature, 437(7057), 436-436 (2005)
Inosha D Gomes et al.
Chembiochem : a European journal of chemical biology, 20(11), 1444-1449 (2019-02-01)
Histone deacetylase 1 (HDAC1) regulates transcription by deacetylating histones. In addition to histones, several non-histone proteins are HDAC1 substrates, which suggests a role for HDAC1 beyond epigenetics. Unfortunately, the identification of non-histone substrates has been largely serendipitous, which makes full characterization
Histone demethylation mediated by the nuclear amine oxidase homolog LSD1
Shi Y, et al.
Cell, 119(7), 941-953 (2004)
Lysine-specific demethylase 1 regulates the embryonic transcriptome and CoREST stability
Foster CT, et al.
Molecular and Cellular Biology, 30(20), 4851-4863 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service