Skip to Content
Merck
All Photos(1)

Key Documents

L7293

Sigma-Aldrich

Monoclonal Anti-LSD1 (AOF2) antibody produced in mouse

~2 mg/mL, clone LSD1-12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Amine oxidase, flavin-containing, 2, Anti-BHC110, Anti-BRAF-HDAC complex protein 110, Anti-KIAA0601 protein, Anti-Lysine-specific demethylase 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LSD1-12, monoclonal

form

buffered aqueous solution

mol wt

antigen ~110 kDa

species reactivity

mouse, human

concentration

~2 mg/mL

technique(s)

western blot: 0.5-1 μg/mL using MCF7 total cell extract

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AOF2(23028)
mouse ... Aof2(99982)

General description

Monoclonal Anti-LSD1 (AOF2) (mouse IgG1 isotype) is derived from the hybridoma LSD1-12 produced by the fusion of mouse myeloma cells (NS1) and splenocytes from BALB/c mice. Lysine specific demethylase 1 (LSD1) also known as amine oxidase flavin-containing domain 2 (AOF2) () is a lysine-specific demethylase. It is a nuclear protein containing a SWIRM (Swi3p, Rsc8p and Moira) domain, a flavin adenine dinucleotide (FAD)-binding motif and an amine oxidase domain. The LSD1 gene is mapped to human chromosome 1p36.12. LSD1 is referred to by additional names including KDM1A protein, BHC110(BRAF-HDAC complex protein 110) and nuclear polyamine oxidase (NPAO)().

Specificity

Monoclonal Anti-LSD1 (AOF2) recognizes human and mouse LSD1.

Immunogen

synthetic peptide corresponding to amino acids 832-847 of human LSD1/BHC110. This sequence is identical in bovine, rat, mouse, and frog.

Application

Monoclonal Anti-LSD1 (AOF2) antibody produced in mouse has been used:
  • immunofluorescence
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry

Biochem/physiol Actions

Lysine Specific Demethylase 1 (LSD1) is a component of several histone deacetylase complexes that function through modifying chromatin structure to repress transcription.LSD1 action can signal either activation or repression of transcription. Demethylation of histone H3 Lys4 by LSD1 leads to transcriptional repression of target genes, while demethylation of histone H3 at Lys9 leads to depression of androgen receptor target genes. The activity of LSD1 (AOF2) is regulated by its associated factors namely, corepressor to the REST (RE1 silencing transcription factor/neural restrictive silencing factor)(CoREST). This, in turn, promotes demethylation by enhancing the association of LSD1 with the nucleosomes and also protects LSD1 from proteasomal degradation. LSD1 is a component of nucleosome remodeling and deacetylase (NuRD). It regulates neuronal development and favors neural stem cell proliferation. It highly expressed in acute myeloid leukemia and many solid tumors.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Wang et al.
Nature genetics, 41(1), 125-129 (2008-12-23)
Histone methylation and DNA methylation cooperatively regulate chromatin structure and gene activity. How these two systems coordinate with each other remains unclear. Here we study the biological function of lysine-specific demethylase 1 (LSD1, also known as KDM1 and AOF2), which
The cancer driver genes IDH1 2, JARID1C KDM5C, and UTX KDM6A: crosstalk between histone demethylation and hypoxic reprogramming in cancer metabolism
Chang S, et al.
Experimental & Molecular Medicine, 51(6), 1-17 (2019)
Daniela Magliulo et al.
Frontiers in oncology, 8, 255-255 (2018-08-04)
Acute myeloid leukemia (AML) is a heterogeneous hematopoietic malignancy characterized by the accumulation of incompletely differentiated progenitor cells (blasts) in the bone marrow and blood, and by suppression of normal hematopoiesis. It has recently become apparent that the AML genome
Alba Maiques-Diaz et al.
Epigenomics, 8(8), 1103-1116 (2016-08-02)
LSD1 (KDM1A; BHC110; AOF2) was the first protein reported to exhibit histone demethylase activity and has since been shown to have multiple essential roles in mammalian biology. Given its enzymatic activity and its high-level expression in many human malignancies, a
Yujiang Shi et al.
Cell, 119(7), 941-953 (2004-12-29)
Posttranslational modifications of histone N-terminal tails impact chromatin structure and gene transcription. While the extent of histone acetylation is determined by both acetyltransferases and deacetylases, it has been unclear whether histone methylation is also regulated by enzymes with opposing activities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service