Skip to Content
Merck
All Photos(1)

Key Documents

L929/A

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

mouse adipose tissue

packaging

tube of 5 μg 14112101-DNA-5UG
pkg of vial of cells 14112101-1VL

growth mode

Adherent

morphology

Fibroblast

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

This Adriamycin-resistant cell line has been developed by exposure of the parent L929 murine fibroblast cell line (ECACC catalogue number 85011425) to increasing concentrations of doxorubicin in-vitro. L929/A cells can be used in the development of novel anti-cancer treatments. Resistance can be circumvented by modulating agents such as verapamil and quinine. The parent cell line L929 was derived from normal subcutaneous areolar adipose tissue.

Culture Medium

RPMI 1640 + 10% Fetal Bovine Serum (FBS) + 2mM L-Glutamine + 0.086 uM Doxorubicin hydrochloride.

Subculture Routine

Trypsinize with 0.05% Trypsin/EDTA and inoculate new cultures vessels at 1-2 x 104 cells/cm2. Cultures incubated in a humidified 5% CO2/95% air incubator at 37oC.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service