Skip to Content
Merck
All Photos(2)

Documents

04693124001

Roche

cOmplete, Mini Protease Inhibitor Cocktail

Tablets provided in EASYpacks

Synonym(s):

protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21

form

tablet

packaging

pack of 30 tablets (individually packed in foil blister packs)

manufacturer/tradename

Roche

solubility

aqueous buffer: soluble
water: soluble

storage temp.

2-8°C

General description

cOmplete, Mini Protease Inhibitor Tablets, inhibit a broad spectrum of serine, cysteine and metalloproteases, as well as calpains. Due to the optimized composition of the tablets, they show excellent inhibition effects, and are well suited for the protection of proteins isolated from animal tissues, plants, yeast, and bacteria.
cOmplete, Mini, contains EDTA and both irreversible and reversible protease inhibitors.

Specificity

For the inhibition of serine, cysteine, and metalloproteases in bacterial, mammalian, yeast, and plant cell extracts.

Application

cOmplete, Mini Protease Inhibitor Cocktail has been used as a component in
  • mammalian-protein extraction reagent (M-PER) for lysis of glioma stem cells (GSCs)
  • sodium dodecyl sulphate-tris buffered saline (SDS-TBS) for the homogenisation of brain tissues
  • radioimmunoprecipitation assay (RIPA) buffer for lysis of human embryonic stem cells (hESCs)

cOmplete, Mini, is used for the inhibition of serine, cysteine, and metalloproteases in bacterial, mammalian, yeast, and plant cell extracts.

cOmplete, Mini, contains both reversible and irreversible protease inhibitors. Therefore, we recommend the addition of cOmplete, Mini, to all stock buffers and solutions normally protected with protease inhibitors, and not  just during the initial purification steps.
cOmplete, Mini, contains EDTA (1 tablet yields a 1 mM EDTA solution in 10 ml). Therefore, the extraction buffer should not contain divalent cations like Ca2+, Mg2+, or Mn2+, otherwise the inhibition of the metalloproteases might be incomplete. If the protein of interest will be purified by IMAC (Immobilized Metal Affinity Chromatography), for example, Poly-His-tagged recombinant proteins, EDTA has to be eliminated (e.g., by dialysis) prior to the chromatography.

Alternatively, the product cOmplete, Mini, EDTA-free, can be used. These tablets are identical to cOmplete, Mini, with the exception that no EDTA or other chelating agent is  present.

cOmplete, Mini Tablets, inhibit proteolytic activity in small extraction volumes (up to 10 ml). If you are working with a larger volume (up to 50 ml), we recommend to use a different cOmplete product. The composition of the cOmplete Tablet is identical to the cOmplete, Mini Tablet, therefore, comparable results are achieved with both product types.

Features and Benefits

  • Convenience: Simply push the quick-dissolving tablet through the foil packaging of the EASYpack into your buffer.
  • Complete protection: Instantly protect your proteins against a broad range of proteases (see Table 1).
  • Flexible: Protect proteins in extracts from almost any tissue or cell, including animals, plants, yeast, bacteria, or fungi.
  • Safe: Choose non-toxic inhibitors that pose no risk to you or those around you.
  • Suitable for IMAC purification: Purify poly-histidine tagged proteins with cOmplete His-Tag Purification Resin.

Packaging

30 individually packed cOmplete Protease Inhibitor Cocktail Tablets in foil blisters. Each tablet is sufficient for a volume of 10 ml solution.

Components

Proprietary mixture of several protease inhibitors with broad inhibitory specificity.

Quality

Performance-tested with pancreas extract.

Physical form

Tablets in foil blister packs

Preparation Note

Working concentration: 1 tablet per 10 ml extraction solution

Preparation of Working Solution:
Carefully push the tablet through the foil packaging using the base of your thumb (not fingernail) to prevent breakage of tablets.

One cOmplete Mini tablet is sufficient for the inhibition of the proteolytic activity in 10 ml extraction solution. When very high proteolytic activity is present, one tablet should be used for 7 ml extraction buffer. The tablets can be added directly to the extraction medium. Alternatively, a stock solution (7 X conc.) can be prepared.

Stock solution (7 X conc.):
Dissolve one cOmplete Mini tablet in 1.5 ml dist. water or in 1.5 ml 100 mM phosphate buffer, pH 7.0.

Storage conditions (working solution): The stock solution can be stored at 2 to 8 °C for 1 to 2 weeks, or at least 12 weeks at -15 to -25 °C.

Reconstitution

Soluble in aqueous buffers, or add directly to extraction media. Alternatively, prepare 7x stock solutions in 1.5 ml water or 100 mM phosphate buffer, pH 7.0. Stock solution is stable for 1-2 weeks at 2 to 8 °C or at least 12 weeks at -15 to -25 °C; can be used in thiol-containing solutions at 15 to 25 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

cOmplete is a trademark of Roche

related product

Pictograms

Exclamation markExclamation mark

Signal Word

Warning-Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of the 18 kda translocator protein (tspo) expression in post-mortem normal and alzheimer's disease brains
Gui Y, et al.
Brain Pathology, 1-14 (2019)
Yu-Shi Wang et al.
Biomolecules, 10(4) (2020-04-05)
(1) Background: Epithelial-mesenchymal transition (EMT) is an essential step for cancer metastasis; targeting EMT is an important path for cancer treatment and drug development. NF-κB, an important transcription factor, has been shown to be responsible for cancer metastasis by enhancing
Jakob Triebel et al.
European journal of endocrinology, 161(2), 345-353 (2009-05-30)
In vitro experiments and in vivo studies on rodents demonstrate that N-terminal 14, 15, 16, 17, and 18 kDa fragments prolactin-related vasoinhibin (PRL-V) of human PRL are natural inhibitors of neovascularization in the retina and elsewhere. These N-terminal PRL fragments
Hila Emmert et al.
Experimental dermatology, 29(8), 749-758 (2020-07-09)
Emerging evidence suggests oxidative stress plays a role in the pathophysiology of both atopic dermatitis (AD) and psoriasis (PSO). We established in vitro models of AD and PSO skin, and characterized these models in regard to their oxidative stress state.
Elizabeth C Barretto et al.
Genetics, 215(4), 1013-1025 (2020-06-10)
Exposure of tissues and organs to low oxygen (hypoxia) occurs in both physiological and pathological conditions in animals. Under these conditions, organisms have to adapt their physiology to ensure proper functioning and survival. Here, we define a role for the

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service