Skip to Content
Merck
All Photos(5)

Key Documents

G1N70

Sigma-Aldrich

GenElute Mammalian Genomic DNA Miniprep Kits

sufficient for 70 purifications

Synonym(s):

Mammalian Genomic DNA Miniprep, animal gDNA prep, gDNA miniprep, mammalian gDNA extraction, mammalian gDNA isolation, mammalian gDNA prep, mammalian gDNA purification, silica mammalian gDNA prep, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52
Pricing and availability is not currently available.

usage

sufficient for 70 purifications

Quality Level

technique(s)

DNA purification: suitable

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute Mammalian Genomic DNA Purification Kit provides a simple and convenient method to isolate pure, high molecular weight DNA from a variety of mammalian sources. These kits use a silica-based membrane, specially selected for genomic DNA purification, in a convenient spin column format.

The starting materials are lysed in a chaotropic salt-containing solution to ensure the thorough denaturation of macromolecules. The addition of ethanol causes DNA to bind when the lysate is spun through a silica membrane in a microcentrifuge tube. After washing to remove the contaminants, the DNA is eluted in 200 μL of a Tris-EDTA solution. The expected yields of genomic DNA will vary depending on the amount and type of starting material used. DNA purified with the kit has an A260/A280 ratio between 1.6 and 1.9 and can be up to 50 kb in length.

The purified genomic DNA is ready for immediate use in downstream applications such as restriction digest, PCR, southern blots, and sequencing reactions.
The GenElute Mammalian Genomic DNA Purification Kit provides a simple and convenient way to isolate pure, high molecular weight DNA from a variety of mammalian sources. These kits use a silica-based membrane, specially selected for genomic DNA purification, in a convenient spin column format.

Isolate pure genomic DNA from cultured cells, tissues (including rodent tails), and fresh whole blood or white blood cells. The kit combines the advantages of silica binding with a microspin format and eliminates the need for expensive resins, alcohol precipitation, and hazardous organic compounds such as phenol and chloroform.

Application

The purified genomic DNA is ready for immediate use in downstream applications such as:
  • restriction endonuclease digestions
  • PCR
  • Southern blots
  • sequencing reactions
  • cloning
  • ligation

Features and Benefits

  • Expected yield: 25 μg from 2 x 106 cultured cells; 30 μg from 25 mg of tissue
  • Elution volume: 200 - 400 μl
  • Time required: 20 min after lysis
  • A260/A280 ratio: 1.6 - 1.9
  • Mechanical homogenization required: No

Principle

The starting materials are lysed in a chaotropic salt-containing solution to ensure the thorough denaturation of macromolecules. Addition of ethanol causes DNA to bind when the lysate is spun through a silica membrane in a microcentrifuge tube. After washing to remove the contaminants, the DNA is eluted in 200 μL of a Tris-EDTA solution. The expected yields of genomic DNA will vary depending on the amount and type of starting material used. DNA purified with the kit has an A260/A280 ratio between 1.6 and 1.9 and can be up to 50 kb in length.

Other Notes

For additional information, please see www.sigma-aldrich.com/genomicdna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • C2112Column Preparation SolutionSDS

  • P2308Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biologySDS

  • R6148RNase A solutionSDS

recommended

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew J Binnington et al.
Pharmacogenetics and genomics, 22(6), 429-440 (2012-05-10)
Alaska Native (AN) people have a high prevalence of tobacco use and associated morbidity and mortality when compared with the general USA population. Variations in the CYP2A6 and CYP2B6 genes, encoding enzymes responsible for nicotine metabolic inactivation and procarcinogen activation
C M Vaz et al.
Acta biomaterialia, 1(5), 575-582 (2006-05-17)
Aiming to develop a scaffold architecture mimicking morphological and mechanically that of a blood vessel, a sequential multi-layering electrospinning (ME) was performed on a rotating mandrel-type collector. A bi-layered tubular scaffold composed of a stiff and oriented PLA outside fibrous
Iris Amitay-Laish et al.
Journal of the American Academy of Dermatology, 66(1), 46-50 (2011-06-28)
Human herpesvirus (HHV) 8, an essential etiologic agent of Kaposi sarcoma, is also associated with several lymphoproliferative disorders. The involvement of HHV 8 in mycosis fungoides (MF) and large plaque parapsoriasis (LPP) is controversial, with contradictory reports from various countries
Justin Tan et al.
Hearing research, 268(1-2), 46-59 (2010-05-15)
In the adult rodent inner ear, p75NTR is weakly expressed in primary auditory neurons (PANs) and cochlear Schwann cells. When the organ of Corti is damaged during trauma, its expression dramatically increases. It is unclear what role p75NTR plays under
O S Petrakova et al.
Acta naturae, 7(4), 80-92 (2016-01-23)
The studies of cell plasticity and differentiation abilities are important problems in modern cellular biology. The use of histone deacetylase inhibitor - valproic acid is a promising approach to increasing the differentiation efficiency of various cell types. In this paper

Protocols

The GenElute Mammalian Genomic DNA Purification Kit Protocol describes the isolation of pure, high molecular weight DNA from a variety of mammalian sources.

The GenElute Mammalian Genomic DNA Purification Kit Protocol describes the isolation of pure, high molecular weight DNA from a variety of mammalian sources.

The GenElute Mammalian Genomic DNA Purification Kit Protocol describes the isolation of pure, high molecular weight DNA from a variety of mammalian sources.

The GenElute Mammalian Genomic DNA Purification Kit Protocol describes the isolation of pure, high molecular weight DNA from a variety of mammalian sources.

Related Content

GenElute Mammalian Genomic DNA Purification Kit isolates pure DNA from mammalian cells using a silica-based membrane in spin columns.

GenElute Mammalian Genomic DNA Purification Kit isolates pure DNA from mammalian cells using a silica-based membrane in spin columns.

GenElute Mammalian Genomic DNA Purification Kit isolates pure DNA from mammalian cells using a silica-based membrane in spin columns.

GenElute Mammalian Genomic DNA Purification Kit isolates pure DNA from mammalian cells using a silica-based membrane in spin columns.

See All

Questions

  1. Will the kit also isolate mitochondrial DNA?

    1 answer
    1. R&D confirms that you will obtain mtDNA in addition to genomic DNA when following a standard protocol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service