Skip to Content
Merck
All Photos(6)

Key Documents

G2N10

Sigma-Aldrich

GenElute Plant Genomic DNA Miniprep Kit

greener alternative

sufficient for 10 purifications

Synonym(s):

Plant Genomic DNA Miniprep, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55

usage

sufficient for 10 purifications

Quality Level

greener alternative product characteristics

Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute Plant Genomic DNA Miniprep Kit provides a simple and convenient way to isolate pure DNA from a variety of plant species. The GenElute kit combines the advantages of a silica-based system with a microspin format and eliminates the need for expensive resins, RNase treatment, and hazardous organic compounds such as phenol and chloroform.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Application

GenElute Plant Genomic DNA Miniprep Kit has been used:
  • in isolation of arbuscular mycorrhizal (AM) fungal spores DNA
  • in the purification of cetyl trimethyl ammonium bromide (CTAB)-extracted genomic DNA
  • in genomic DNA isolation from leaves

The purified genomic DNA is ready for immediate use in sensitive downstream applications such as:
  • PCR
  • restriction endonuclease digestion
  • cloning
  • Southern blots

Features and Benefits

  • Starting material: Up to 100 mg of plant tissue
  • Expected yield: Up to 20 μg
  • Elution volume: 100 - 200 μl
  • Time required: < 40 min
  • RNase treatment required: No

Principle

Several micrograms of DNA can be obtained from up to 100 mg of fresh tissue or 10 mg of freeze-dried material in less than an hour. Plant tissue is disrupted by grinding in liquid nitrogen and the DNA is released with detergent and chaotrope. Proteins, polysaccharides, and cell debris are eliminated with a 10 minute precipitation procedure followed by centrifugation through a filtration column, included in the kit. The genomic DNA is purified further by a silica bind-wash-elute procedure in microcentrifuge spin columns. The purified DNA is greater than 20 kb in length.

Other Notes

For additional information, please see www.sigma-aldrich.com/genomicdna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • C2112Column Preparation SolutionSDS

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phytohormone interplay controls proliferation of in vitro cultivated cells of Arabidopsis thaliana ethylene-insensitive mutants.
N S Stepanchenko et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 442, 46-49 (2012-03-20)
Glomus hyderabadensis, a new species: its taxonomy and phylogenetic comparison with related species
Rani SS, et al.
Mycotaxon, 89(2), 245-254 (2004)
Ellen Erzeel et al.
Plant molecular biology, 81(4-5), 401-415 (2013-01-19)
Lysine biosynthesis in plants is tightly regulated by feedback inhibition of the end product on the first enzyme of the lysine-specific branch, dihydrodipicolinate synthase (DHDPS). Three complete DHDPS coding sequences and one partial sequence were obtained in Medicago truncatula via
Peter Kämpfer et al.
International journal of systematic and evolutionary microbiology, 62(Pt 6), 1342-1347 (2011-08-11)
The taxonomy of strain CCUG 55240(T), a Gram-staining-positive, aerobic, endospore-forming bacterium that was isolated from a paper mill, was investigated using a polyphasic approach. In phylogenetic analysis based on 16S rRNA gene sequences, the novel strain was grouped with established
Stefanie P Glaeser et al.
International journal of systematic and evolutionary microbiology, 63(Pt 2), 521-525 (2012-04-24)
A Gram-staining-negative, rod-shaped, non-spore-forming bacterium, designated strain E103(T), was isolated from the skin of the medical leech Hirudo verbana. 16S rRNA gene sequence analysis showed that the isolate was closely related to species of the genus Castellaniella. Castellaniella ginsengisoli DCY36(T)

Protocols

This protocol describes a simple and convenient procedure to isolate pure DNA from a variety of plant species using the GenElute Plant Genomic DNA Miniprep Kit.

GenomePlex® Whole Genome Amplification has been used to amplify genomic DNA from soybean, corn, tomato, purple coneflower, and ginseng.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service