Skip to Content
Merck
All Photos(4)

Key Documents

NA2010

Sigma-Aldrich

GenElute Blood Genomic DNA Kit

sufficient for 70 purifications

Synonym(s):

Blood Genomic DNA, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55

usage

sufficient for 70 purifications

technique(s)

DNA purification: suitable

storage temp.

15-25°C
room temp

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute Blood Genomic DNA kit provides a simple and convenient way to isolate pure genomic DNA from fresh or aged (older than 24 hours) whole blood. The kit combines the advantages of silica binding with a microspin format, and eliminates the need for expensive resins, alcohol precipitation, and hazardous organic compounds such as phenol and chloroform.

Application

GenElute Blood Genomic DNA Kit has been used:
  • to extract DNA from samples of large granules (LG) and smaller granules (SG) obtained from vasectomized rabbits
  • to isolate DNA from the blood samples 
  • to extract whole blood DNA from the left blood cells

The purified genomic DNA is ready for downstream applications such as:
  • restriction endonuclease digestions
  • PCR
  • Southern blots
  • sequencing reactions
  • cloning

Features and Benefits

  • Starting material: Up to 200 μl of fresh or aged blood
  • Expected yield: Up to 10 μg
  • Elution volume: 400 μl
  • Time required: <40 min
  • A260/A280 ratio: 1.6 - 1.9
  • Compatible with many anticoagulants, including EDTA, Heparin, and Sodium Citrate
  • Typical yields 4-10 μg from 200 μL of whole blood
  • Pure genomic DNA with a A260/A280 ratio between 1.6 and 1.9
  • Isolate DNA up to 50 kb in length

Principle

The starting material is lysed in a chaotropic salt-containing solution to ensure the thorough denaturation of macromolecules. The addition of ethanol causes the DNA to bind when the lysate is spun through a silica membrane in a microcentrifuge tube. A Prewash Solution is provided to help remove contaminants that are associated with aged (older than 24 hours) whole blood samples. After washing to remove contaminants, the DNA is eluted in 200 mL of a Tris-EDTA solution.

The expected yields of genomic DNA will vary depending on the amount and nature of the starting material used (for example, 4 to 10 μg of RNase A-treated DNA can be isolated from 200 μl of fresh whole blood in less than one hour). DNA purified with this kit has an A260/A280 ratio between 1.6 and 1.9 and can be up to 50 kb in length.

Other Notes

For additional information, please see www.sigma-aldrich.com/genomicdna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • C2112Column Preparation SolutionSDS

  • P2308Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biologySDS

  • R6148RNase A solutionSDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sai Wang et al.
Frontiers in genetics, 11, 585064-585064 (2020-12-01)
Familial renal glucosuria is a rare renal tubular disorder caused by SLC5A2 gene variants. Most of them are exonic variants and have been classified as missense variants. However, there is growing evidence that some of these variants can be detrimental
Xiangzhong Zhao et al.
Scientific reports, 6, 33920-33920 (2016-09-27)
Familial renal glycosuria (FRG) is caused by mutations in the SLC5A2 gene, which codes for Na+-glucose co-transporters 2 (SGLT2). The aim of this study was to analyze and identify the mutations in 16 patients from 8 families with FRG. All
Yanxia Gao et al.
Renal failure, 36(8), 1226-1232 (2014-07-01)
The objective of this study is to identify ATP6V1B1, ATP6V0A4 and SLC4A1 genes mutations and assess audiologic characteristics in six Chinese children with primary distal renal tubular acidosis from four unrelated families between the ages of 2 and 13 years.
Cui Wang et al.
Scientific reports, 6, 33652-33652 (2016-09-21)
Primary hyperoxaluria type 1 (PH1) is a rare genetic disease characterized by excessive oxalate accumulation in plasma and urine, resulting in various phenotypes because of allelic and clinical heterogeneity. This study aimed to detect disease-associated genetic mutations in three PH1
Xinsheng Wang et al.
American journal of nephrology, 42(1), 78-84 (2015-09-05)
Twenty-six HOGA1 mutations have been reported in primary hyperoxaluria (PH) type 3 (PH3) patients with c.700 + 5G>T accounting for about 50% of the total alleles. However, PH3 has never been described in Asians. A Chinese child with early-onset nephrolithiasis

Protocols

Whole genome amplification (WGA) of plasma and serum DNA presents a unique challenge due to the small amount of nucleic acid in such samples.

Whole genome amplification (WGA) of plasma and serum DNA presents a unique challenge due to the small amount of nucleic acid in such samples.

Whole genome amplification (WGA) of plasma and serum DNA presents a unique challenge due to the small amount of nucleic acid in such samples.

Whole genome amplification (WGA) of plasma and serum DNA presents a unique challenge due to the small amount of nucleic acid in such samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service