Skip to Content
Merck
All Photos(3)

Documents

A9434

Sigma-Aldrich

Ammonium chloride

for molecular biology, suitable for cell culture, ≥99.5%

Synonym(s):

Salmiac

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
Beilstein:
4371014
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050207
PubChem Substance ID:
NACRES:
NA.31

grade

for molecular biology

vapor density

1.9 (vs air)

vapor pressure

1 mmHg ( 160.4 °C)

Assay

≥99.5%

form

powder or crystals

technique(s)

cell culture | mammalian: suitable

mp

340 °C (subl.) (lit.)

anion traces

phosphate (PO43-): ≤2 ppm
sulfate (SO42-): ≤20 ppm

cation traces

Fe: ≤2 ppm
heavy metals (as Pb): ≤5 ppm

foreign activity

DNase, RNase, protease, none detected

SMILES string

N.Cl

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium chloride has been used:
  • as an inhibitor of endosome acidification in vero cells
  • as an autophagy inhibitor in vascular smooth muscle cell (VSMC)
  • in the depletion and lysis of erythrocytes in testis parenchyma homogenate

Biochem/physiol Actions

Ammonium chloride is one of the three principal components of the nitrogen cycle. Ammonium chloride administration promotes creatinine and urea clearance andinduces metabolic acidosis in mice. Excess of ammonium chloride inhibits Kreb′s pathway in brain and depletes ATP in astrocytes. Ammonium chloride inhibits acidification in the endosome-lysosome system. Ammonium chloride is a hemolysing agent used for the lysis of erythrocytes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aqueous ammonia and ammonium chloride hydrates: Principal infrared spectra
Max JJ and Chapados C
Journal of Molecular Structure, 1046, 124-135 (2013)
Effect of ammonium chloride and dietary phosphorus in the azotaemic rat. I. Renal function and biochemical changes
Jara A, et al.
Nephrology, Dialysis, and Transplantation, 19(8), 1986-1992 (2004)
Infectious bronchitis virus entry mainly depends on clathrin mediated endocytosis and requires classical endosomal/lysosomal system
Wang H, et al.
Virology, 528, 118-136 (2019)
Erythrocyte lysis in isotonic solution of ammonium chloride: Theoretical modeling and experimental verification
Chernyshev AV, et al.
Journal of Theoretical Biology, 251(1), 93-107 (2008)
Nrf2 Activation Provides Atheroprotection in Diabetic Mice Through Concerted Upregulation of Antioxidant, Anti-inflammatory, and Autophagy Mechanisms
Lazaro I, et al.
Frontiers in Pharmacology, 9, 118-136 (2018)

Protocols

The GenElute Mammalian Genomic DNA Purification Kit Protocol describes the isolation of pure, high molecular weight DNA from a variety of mammalian sources.

The GenElute Mammalian Genomic DNA Purification Kit Protocol describes the isolation of pure, high molecular weight DNA from a variety of mammalian sources.

The GenElute Mammalian Genomic DNA Purification Kit Protocol describes the isolation of pure, high molecular weight DNA from a variety of mammalian sources.

The GenElute Mammalian Genomic DNA Purification Kit Protocol describes the isolation of pure, high molecular weight DNA from a variety of mammalian sources.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service