Skip to Content
Merck
All Photos(6)

Key Documents

G2N350

Sigma-Aldrich

GenElute Plant Genomic DNA Miniprep Kit

greener alternative

sufficient for 350 purifications

Synonym(s):

Plant Genomic DNA Miniprep, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55

usage

sufficient for 350 purifications

Quality Level

greener alternative product characteristics

Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

DNA purification: suitable

greener alternative category

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute Plant Genomic DNA Miniprep Kit provides a simple and convenient way to isolate pure DNA from a variety of plant species. The GenElute kit combines the advantages of a silica-based system with a microspin format and eliminates the need for expensive resins, RNase treatment, and hazardous organic compounds such as phenol and chloroform.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Application

The purified genomic DNA is ready for immediate use in sensitive downstream applications such as:
  • PCR
  • restriction endonuclease digestion
  • cloning
  • Southern blots

Features and Benefits

  • Starting material: Up to 100 mg of plant tissue
  • Expected yield: Up to 20 μg
  • Elution volume: 100 - 200 μl
  • Time required: < 40 min
  • RNase treatment required: No

Principle

Several micrograms of DNA can be obtained from up to 100 mg of fresh tissue or 10 mg of freeze-dried material in less than an hour. Plant tissue is disrupted by grinding in liquid nitrogen and the DNA is released with detergent and chaotrope. Proteins, polysaccharides, and cell debris are eliminated with a 10 minute precipitation procedure followed by centrifugation through a filtration column, included in the kit. The genomic DNA is purified further by a silica bind-wash-elute procedure in microcentrifuge spin columns. The purified DNA is greater than 20 kb in length.

Other Notes

For additional information, please see www.sigma-aldrich.com/genomicdna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • C2112Column Preparation SolutionSDS

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Lunn et al.
Plant physiology and biochemistry : PPB, 66, 91-97 (2013-03-19)
Fruit development entails a multitude of biochemical changes leading up to the mature green stage. During this period the cell wall will undergo complex compositional and structural changes. Inhibition of genes encoding elements of the machinery involved in trafficking to
V Roussel et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 111(1), 162-170 (2005-05-12)
A sample of 480 bread wheat varieties originating from 15 European geographical areas and released from 1840 to 2000 were analysed with a set of 39 microsatellite markers. The total number of alleles ranged from 4 to 40, with an
Catherine Ravel et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 112(4), 738-743 (2005-12-20)
A previous study in wheat (Triticum aestivum L.) identified two candidate genes controlling a quantitative trait locus (QTL) for high-molecular-weight glutenin subunit (HMW-GS) GluBx. These candidates were Glu-B1-1, the structural gene coding for Glu1Bx, and the B homoeologous gene coding
V Roussel et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 108(5), 920-930 (2003-11-14)
A set of 41 wheat microsatellite markers (WMS), giving 42 polymorphic loci (two loci on each chromosome), was used to describe genetic diversity in a sample of 559 French bread wheat accessions (landraces and registered varieties) cultivated between 1800 and
Minal Mhatre
Methods in molecular biology (Clifton, N.J.), 11013, 435-453 (2012-11-28)
Pineapple (Ananas comosus L., Merr.) is a commercially important crop, grown in the tropical and subtropical regions. However, the crop is faced with postharvest damage and poor varietal and nutritional improvement. Being a vegetatively propagated crop, conventional breeding programs take

Protocols

This protocol describes a simple and convenient procedure to isolate pure DNA from a variety of plant species using the GenElute Plant Genomic DNA Miniprep Kit.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service