Skip to Content
Merck
All Photos(2)

Documents

A5795

Sigma-Aldrich

Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

HRP Rabbit Anti-Rat IgG (whole molecule)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

should not react with

human

technique(s)

direct ELISA: 1:50,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:750
western blot (chemiluminescent): 1:80,000-1:160,000 using rat PC12 cell extract

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

An immunoglobulin has two heavy chain and two light chain connected by disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Rat has five immunoglobulin classes- IgM, IgG, IgA, IgD and IgGE. IgG has four subclasses- IgG1, IgG2a, IgG2b and IgG2c.
IgGs are glycoprotein antibodies that modulate several immune responses. Rat IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rat IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins.
Anti-Rat IgG (whole molecule)-Peroxidase antibody binds all rat Igs and does not react with human IgG.

Immunogen

purified rat IgG

Application

Anti-Rat IgG (whole molecule)-Peroxidase antibody has been used as a secondary antibody in immunohistochemical applications at a dilution of 1:1500. The product has also been used for western blot analysis at 1:20,000. Furthermore, the antibody can be used for direct ELISA at 1:50,000.
Anti-Rat IgG (whole molecule)?Peroxidase antibody produced in rabbit has been used in immunohistochemistry and western blot analysis.
HA-tagged proteins over expressed in S. cerevisiae were detected by western blot using HRP-conjugated rabbit anti-rat IgG as the secondary antibody.

Biochem/physiol Actions

Immunoglobulin (IgG) helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity. IgG participates in hypersensitivity type II and type III.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT

Preparation Note

Adsorbed with human IgG to reduce background staining with human samples.
Prepared by two-step glutaraldehyde method.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Bensmihen et al.
FEBS letters, 561(1-3), 127-131 (2004-03-12)
The Arabidopsis abscisic acid (ABA) insensitive (ABI)5 transcription factor participates in the ABA-dependent induction of late embryogenesis abundant (LEA) genes in the final stages of seed development. We tested whether the VP16 transcriptional activation domain is sufficient to provide ABI5
Galectin-3 as a potential therapeutic target in tumors arising from malignant endothelia
Johnson KD, et al.
Neoplasia, 9(8), 662-662 (2007)
Kim D Johnson et al.
Neoplasia (New York, N.Y.), 9(8), 662-670 (2007-09-06)
Angiosarcoma (ASA) in humans and hemangiosarcoma (HSA) in dogs are deadly neoplastic diseases characterized by an aggressive growth of malignant cells with endothelial phenotype, widespread metastasis, and poor response to chemotherapy. Galectin-3 (Gal-3), a beta-galactoside-binding lectin implicated in tumor progression
Probing the Function of STAS Domains of the Arabidopsis Sulfate Transporters.
Shibagaki, N.
Journal of Biochemistry, 279, 30791-30799 (2004)
Medhat A Al-Ghobashy et al.
Scientific reports, 7, 46468-46468 (2017-04-21)
Recombinant human myelin basic protein (rhMBP) was previously produced in the milk of transgenic cows. Differences in molecular recognition of either hMBP or rhMBP by surface-immobilized anti-hMBP antibodies were demonstrated. This indicated differences in immunological response between rhMBP and hMBP.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service