Skip to Content
Merck
All Photos(1)

Key Documents

12013819001

Roche

Anti-HA-Peroxidase, High Affinity

from rat IgG1

Synonym(s):

antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rat

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

clone 3F10, monoclonal

form

lyophilized (clear, colorless solution after reconstitution)

packaging

pkg of 25 U (25 μg)

manufacturer/tradename

Roche

isotype

IgG1

epitope sequence

YPYDVPDYA

storage temp.

2-8°C

Related Categories

General description

Anti-HA-Peroxidase, High Affinity is a monoclonal antibody to the HA-peptide (clone 3F10), conjugated to peroxidase.

Specificity

Anti-HA-Peroxidase, High Affinity (3F10) recognizes the 9-amino acid sequence YPYDVPDYA, derived from the human influenza hemagglutinin (HA) protein.
This epitope is also recognized in fusion proteins regardless of its position (N-terminal, C-terminal or internal).

Immunogen

The epitope consists of amino acids 98-106 from the human influenza virus hemagglutinin protein.

Application

  • Use Anti-HA-Peroxidase, High Affinity for the detection of HA-tagged recombinant proteins using: ELISA
  • Western blot[1][2]
Since Anti-HA High Affinity is a rat monoclonal, it is possible to use it in conjunction with murine monoclonals for double labeling.

Preparation Note

Stabilizers: proteinaceous stabilizers
Working concentration: The working concentration of conjugate depends on application and substrate.
The following concentrations should be taken as a guideline:
  • Dot blot: 50 mU/ml
  • ELISA: 25 mU/ml
  • Western blot: 50 mU/ml

Reconstitution

Add 1.0 ml double-distilled water for a final concentration of 25 U/mL.
Rehydrate for 10 min prior to use.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranran Wang et al.
Cancers, 12(6) (2020-06-25)
The epigenetic reader BRD4 binds acetylated histones and plays a central role in controlling cellular gene transcription and proliferation. Dysregulation of BRD4's activity has been implicated in the pathogenesis of a wide variety of cancers. While blocking BRD4 interaction with
Julia K Günther et al.
Biology, 9(5) (2020-05-21)
Rigosertib, via reactive oxygen species (ROS), stimulates cJun N-terminal kinases 1/2 (JNK1/2), which inactivate RAS/RAF signaling and thereby inhibit growth and survival of tumor cells. JNK1/2 are not only regulated by ROS-they in turn can also control ROS production. The
Regina Gratz et al.
The New phytologist, 225(1), 250-267 (2019-09-06)
The key basic helix-loop-helix (bHLH) transcription factor in iron (Fe) uptake, FER-LIKE IRON DEFICIENCY-INDUCED TRANSCRIPTION FACTOR (FIT), is controlled by multiple signaling pathways, important to adjust Fe acquisition to growth and environmental constraints. FIT protein exists in active and inactive
Alicja M Cygan et al.
mSphere, 5(1) (2020-02-23)
Toxoplasma gondii is a ubiquitous, intracellular protozoan that extensively modifies infected host cells through secreted effector proteins. Many such effectors must be translocated across the parasitophorous vacuole (PV), in which the parasites replicate, ultimately ending up in the host cytosol
Lina Herhaus et al.
Nature communications, 4, 2519-2519 (2013-09-28)
SMAD transcription factors are key intracellular transducers of TGFβ cytokines. SMADs are tightly regulated to ensure balanced cellular responses to TGFβ signals. Ubiquitylation has a key role in regulating SMAD stability and activity. Several E3 ubiquitin ligases that regulate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service