Skip to Content
Merck
All Photos(1)

Key Documents

H6533

Sigma-Aldrich

Anti-HA−Peroxidase antibody, Mouse monoclonal

clone HA-7, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-HA, Anti-HA, Anti-Influenza Hemagglutinin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HA-7, monoclonal

form

lyophilized powder

packaging

vial of 0.5 mL

concentration

5-11 mg/mL

technique(s)

western blot: 1:4,000 using using HA tagged fusion protein expressed in bacteria, and ECL immunoblotting detection reagent

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Influenza A virus, a RNA virus, encodes two surface glycoproteins, hemagglutinin (HA) and neuraminidase (NA). HA stalk domain based epitope region antibody is effective in neutralizing divergent influenza virus strains.
The mouse monoclonal Anti-HA-Peroxidase antibody recognizes native and denatured-reduced forms of the HA-tagged proteins. It also reacts with the N- or C-terminal HA-tagged fusion proteins expressed in E. coli and mammalian cells.

Specificity

The antibody recognizes native as well as denatured-reduced forms of HA-tagged proteins and is reactive with N- or C-terminal HA-tagged fusion proteins expressed in E. coli or in mammalian cells.

Immunogen

synthetic peptide corresponding to a fragment of human influenza virus hemagglutinin (HA) known as HA-tag, conjugated to KLH

Application

Monoclonal Anti-HA-Peroxidase antibody produced in mouse has been used:
  • in immunopreciptation and immunostaining of human member 9 of solute carrier family 38 SLC38A9 transfected in HeLa cells
  • in immunoblot detection in transfected A375 cells
  • is suitable for immunoblotting of nucleosome assembly protein NAP1L1

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% MIT.

Preparation Note

Reconstitute in .5mL of distilled water.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulina Bartuzi et al.
Nature communications, 7, 10961-10961 (2016-03-12)
The low-density lipoprotein receptor (LDLR) plays a pivotal role in clearing atherogenic circulating low-density lipoprotein (LDL) cholesterol. Here we show that the COMMD/CCDC22/CCDC93 (CCC) and the Wiskott-Aldrich syndrome protein and SCAR homologue (WASH) complexes are both crucial for endosomal sorting
Ernest C So et al.
The Journal of biological chemistry, 291(11), 5832-5843 (2016-01-13)
Legionella pneumophila, the causative agent of Legionnaire's disease, uses its type IV secretion system to translocate over 300 effector proteins into host cells. These effectors subvert host cell signaling pathways to ensure bacterial proliferation. Despite their importance for pathogenesis, the
Influenza virus hemagglutinin stalk-based antibodies and vaccines
Krammer F and Palese P
Current Opinion in Virology, 3(5), 521-530 (2013)
A broadly reactive human anti-hemagglutinin stem monoclonal antibody that inhibits influenza a virus particle release
Yamayoshi S, et al.
EBioMedicine, 17, 182-191 (2017)
Joanna F McGouran et al.
Chemistry & biology, 20(12), 1447-1455 (2013-12-03)
Posttranslational modification with ubiquitin (Ub) controls many cellular processes, and aberrant ubiquitination can contribute to cancer, immunopathology, and neurodegeneration. The versatility arises from the ability of Ub to form polymer chains with eight distinct linkages via lysine side chains and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service