Skip to Content
Merck
All Photos(3)

Key Documents

F1763

Sigma-Aldrich

Anti-Rat IgG (whole molecule)–FITC antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Rat IgG (whole molecule)–Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:320
indirect immunofluorescence: 1:320

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Anti-Rat IgG (whole molecule) is produced in rabbit using purified rat IgG as the immunogen.
Rat IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in rat serum. Fluorescein isothiocyanate (FITC) is a fluorescein derivative (fluorochrome) used to tag antibodies, including secondary antibodies, for use in fluorescence-based assays and procedures. FITC excites at 495 nm and emits at 521 nm.
Rat IgGs have four subclasses, namely, IgG1, IgG2a, IgG2b, and IgG2c. Rat IgGs against target proteins are often used as primary antibodies in various research applications. Anti-Rat IgG (whole molecule)-FITC antibody binds to all rat immunoglobulins.

Specificity

Rabbit polyclonal anti-Rat IgG (whole molecule)−FITC antibody reacts with rat IgG in vitro, in rat serum and biological fluids.

Immunogen

Purified rat IgG

Application

Anti-Rat IgG (whole molecule)-FITC antibody has been used:
  • in immunolabelling for confocal laser scanning microscopy (CLSM)
  • in immunostaining

Anti-Rat IgG (whole molecule)-FITC antibody is suitable for use in flow cytometry (1:100) and immunofluorescence.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Flow cytometry/Cell sorting (1 paper)
Immunofluorescence (1 paper)
Rabbit polyclonal anti-Rat IgG (whole molecule)-FITC antibody may be used to detect and quantitate the level of IgG in rat serum and biological fluids by fluorescent techniques. It may also be used as a secondary antibody in assays that use pig IgG as the primary antibody.

Biochem/physiol Actions

IgGs are glycoprotein antibodies that modulate several immune responses. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection. Thus, secondary anti-rat IgG is a useful tool for the analysis of target proteins.

Other Notes

Antibody adsorbed with human IgG.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural network of arabinogalactan proteins (AGPs) and pectins in apple fruit during ripening and senescence processes
Leszczuk A, et al.
Plant Science, 275, 36-48 (2018)
Yang Zhou et al.
Experimental and therapeutic medicine, 12(4), 2671-2680 (2016-10-05)
Traditional medicinal plants are widely used as immunomodulatory medicines that help improve health. A total of 50 different plants used for the treatment of toxicity were screened for their in vivo protective effects. Flies were fed a standard cornmeal-yeast medium
Identification of the protective effects of traditional medicinal plants against SDS-induced Drosophila gut damage
Zhou Y, et al.
Experimental and Therapeutic Medicine, 12(4), 2671-2680 (2016)
C H Yeung et al.
Molecular human reproduction, 7(7), 617-624 (2001-06-23)
HE5 (CD52) is a glycoprotein which is secreted by the epididymis and which becomes inserted onto maturing spermatozoa. We have previously shown that, in cynomolgus monkey spermatozoa, changes occur upon maturation rendering cryptic the epitope to the monoclonal antibody CAMPATH-1G;
Sílvia Coimbra et al.
Journal of experimental botany, 58(15-16), 4027-4035 (2007-11-28)
Some of the most important changes that occur in plants during sexual reproduction involve the transition from a sporophytic to a gametophytic type of development. In this paper, these changes were evaluated for Arabidopsis thaliana. The results obtained clearly show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service