Skip to Content
Merck
All Photos(1)

Key Documents

84429

Sigma-Aldrich

Hydrochloric acid

~6 M in H2O, for amino acid analysis

Synonym(s):

Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.21

Product Name

Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis

grade

for amino acid analysis

Quality Level

vapor pressure

190 mmHg ( 25 °C)

form

liquid

quality

ampule

analyte chemical class(es)

amino acids

concentration

5900-6100 mmol/L (alkalimetric)
~6 M in H2O

pH

<1

bp

100 °C (lit.)

solubility

water: miscible

density

1.050-1.150 g/cm3

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The Product is 6M solution of hydrochloric acid in water. Hydrochloric acid is a strong mineral acid. It participates in the synthesis of alloxan (hexahydropyrimidine-2,4,5,6-tetrone) and hydrated aluminum chloride.

Application

Hydrochloric acid may be used in the following studies:
  • Preparation of orcinolhydrochloric acid (Bial′s reagent), which is useful for the quantitative determination of sialic acids.
  • Hydrolytic decomposition of chitin.
  • Preparation of ammonium chloride.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Eagleson M.
Concise Encyclopedia Chemistry, 46-46 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 52-52 (1994)
THE HYDROLYSIS OF CHITIN BY CONCENTRATED HYDROCHLORIC ACID, AND THE PREPARATION OF LOW-MOLECULAR-WEIGHT SUBSTRATES FOR LYSOZYME.
J A RUPLEY
Biochimica et biophysica acta, 83, 245-255 (1964-11-01)
Eagleson M.
Concise Encyclopedia Chemistry, 65-65 (1994)
Quantitative estimation of sialic acids. II. A colorimetric resorcinol-hydrochloric acid method.
L SVENNERHOLM
Biochimica et biophysica acta, 24(3), 604-611 (1957-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service