Skip to Content
Merck

Skip To

2104

Hydrochloric acid

c(HCl) = 0.1 mol/l (0.1N), endotoxin free

Synonym(s):

Hydrogen chloride solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Concentration:
0.1 M HCl (0.1N)
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free

vapor pressure

190 mmHg ( 25 °C)

Quality Level

form

liquid

quality

endotoxin free, volumetric

concentration

0.1 M HCl (0.1N)

pH

<1

bp

100 °C (lit.)

density

1.2 g/cm3 at 25 °C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Application


  • sym-Trisubstituted 1,3,5-Triazine Derivatives as Promising Organic Corrosion Inhibitors for Steel in Acidic Solution.: This study explores the use of trisubstituted triazine derivatives in hydrochloric acid solutions to inhibit steel corrosion. The findings highlight the potential of these compounds in industrial applications where corrosion prevention is critical (El-Faham et al., 2016).

  • Melt-Extruded Eudragit FS-Based Granules for Colonic Drug Delivery.: This research examines the development of drug delivery systems using hydrochloric acid solutions to process granules for targeted drug release in the colon, demonstrating a novel approach to pharmaceutical formulations (Zhang, 2016).

  • Effect of molecular structure of aniline-formaldehyde copolymers on corrosion inhibition of mild steel in hydrochloric acid solution.: The study investigates how different molecular structures of aniline-formaldehyde copolymers affect their efficiency as corrosion inhibitors in hydrochloric acid solutions, providing insights for materials science and industrial applications (Zhang et al., 2015).

  • Preparation and characterization of new solid-phase microextraction fibers obtained by sol-gel technology and zirconium oxide electrodeposited on NiTi alloy.: This paper details the creation and testing of new solid-phase microextraction fibers using hydrochloric acid in the preparation process, highlighting advancements in analytical chemistry techniques (Budziak et al., 2008).

  • Polyaniline-uricase biosensor prepared with template process.: The research describes the fabrication of a polyaniline-uricase biosensor using hydrochloric acid in the template process, offering new possibilities for biosensor technology and its applications in medical diagnostics (Kan et al., 2004).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.098711.602381.60327
concentration

0.1 M HCl (0.1N)

concentration

-

concentration

0.01 M HCl (0.01 N)

concentration

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

form

liquid

form

-

form

liquid

form

liquid

pH

<1

pH

0.6 (20 °C in H2O)

pH

-

pH

-

vapor pressure

190 mmHg ( 25 °C)

vapor pressure

-

vapor pressure

-

vapor pressure

-

bp

100 °C (lit.)

bp

-

bp

-

bp

-


Still not finding the right product?

Explore all of our products under Hydrochloric acid solution


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Marta Kolodziejczak et al.
ACS chemical neuroscience, 6(7), 1219-1230 (2015-04-11)
Maturation of functional neuronal circuits during central nervous system development relies on sophisticated mechanisms. First, axonal and dendritic growth should reach appropriate targets for correct synapse elaboration. Second, pruning and neuronal death are required to eliminate redundant or inappropriate neuronal
Douglas M McMillan et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(7), 1804-1812 (2015-01-30)
CYP2D metabolically activates codeine to morphine, which is required for codeine analgesia. Permeability across the blood-brain barrier, and active efflux, suggests that initial morphine in the brain after codeine is due to brain CYP2D metabolism. Human CYP2D is higher in
Davide R Ceratti et al.
Advanced materials (Deerfield Beach, Fla.), 27(34), 4958-4962 (2015-07-23)
A new, simple bi-phasic dip-coating method is developed. This method is considered as a great improvement of the technique for research, development and production, since expensive, rare, harmful, or time-evolving solutions can now be easily deposited on large surfaces and



Global Trade Item Number

SKUGTIN
B75956-5G04061833441992
2104-50ML04061837484407

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters