Skip to Content
Merck
All Photos(1)

Key Documents

83931

Sigma-Aldrich

RNase AWAY®

decontamination reagent for RNase

Synonym(s):

Surface Decontaminant

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.52

Quality Level

form

liquid

density

1.000 g/mL at 20 °C

storage temp.

room temp

General description

RNase AWAY® is a ready-to-use surfactant that removes RNA and RNases from lab equipment. Suitable for PCR equipment, plastic, glassware, and stainless steel lab surfaces.

Application

RNase AWAY® has been used:
  • in flow cytometry analysis
  • to sterilize coated microstir bars for the formation of colloidal emulsion
  • to remove RNAse from biosafety cabinet and lab equipment
  • as a chemical to analyse the dynamic transcriptional and epigenetic remodeling during astrogliogenesis

Caution

Gloves should be worn when handling this product. RNase Decontamination Reagent is alkaline in nature and will cause irritation if prolonged contact with the skin is allowed. In case of contact with eyes, immediately flush with water for fifteen minutes and contact a physician. If swallowed, do not induce vomiting. Give plenty of water and contact a physician immediately.

Other Notes

Do not use RNase Decontamination Reagent on aluminium, soft metal or gaskets and seals. This product is not intended as reactant. Do not add to reaction.

Legal Information

AWAY is a registered trademark of Molecular BioProducts, Inc.
RNase AWAY is a registered trademark of Molecular Bio-Products, Inc.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stage-Specific Transcription Factors Drive Astrogliogenesis by Remodeling Gene Regulatory Landscapes
Tiwari N, et al.
Cell Stem Cell, 23(4), 557-571 (2018)
A CRISPR-Cas9 Genome Engineering Platform in Primary CD4+ T Cells for the Interrogation of HIV Host Factors
Hultquist JF, et al.
bioRxiv, 205500-205500 (2017)
Mie Scatter and Interfacial Tension Based Real-Time Quantification of Colloidal Emulsion Nucleic Acid Amplification
Nicolini AM, et al.
Advanced Biosystems, 1(10), 1700098-1700098 (2017)
[Importance of the clinico-radiologic diagnosis of cecal diverticulitis].
F Gimeno Solsona et al.
Revista espanola de enfermedades digestivas : organo oficial de la Sociedad Espanola de Patologia Digestiva, 77(2), 166-167 (1990-02-01)
Ward De Spiegelaere et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 58(12), 1045-1056 (2010-08-18)
The development and growth of renal glomeruli is regulated by specific angiogenic growth factors, including vascular endothelial growth factor (VEGF) and the angiopoietins (ANGPT1 and ANGPT2). The expression of these factors has already been studied during metanephric glomerulogenesis, but it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service