Skip to Content
Merck
All Photos(1)

Key Documents

H1758

Sigma-Aldrich

Hydrochloric acid

36.5-38.0%, BioReagent, for molecular biology

Synonym(s):

HCL, Hydrogen Chloride Solution, Muriatic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.31

grade

for molecular biology

Quality Level

vapor density

1.3 (vs air)

vapor pressure

3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)

product line

BioReagent

Assay

36.5-38.0%

form

liquid

concentration

30-50%

color

light yellow

pH

<1 (20 °C)

bp

>100 °C (lit.)

solubility

H2O: soluble

density

1.070 g/cm3 at 20 °C (lit.)

cation traces

Fe: ≤0.2 ppm
heavy metals (as Pb): ≤1 ppm

suitability

suitable for molecular biology

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrochloric acid has been used:
  • for the solubilization of formazan crystals
  • to adjust pH in various buffers
  • for the preparation of giant reed extract

Hydrochloric acid has been used:
  • in cell viability assays
  • for DNA denaturation
  • for pH adjustments in sucrose hydrolysis experiment

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Highly synchronized expression of lineage-specific genes during in vitro hepatic differentiation of human pluripotent stem cell lines
Ghosheh N, et al.
Stem Cells International, 2016 (2016)
Growth-suppressing and algicidal properties of an extract from Arundo donax, an invasive riparian plant, against Prymnesium parvum, an invasive harmful alga
Patino R, et al.
Harmful Algae, 71, 1-9 (2018)
Registration and Alignment Between in vivo Functional and Cytoarchitectonic Maps of Mouse Visual Cortex
Zhuang J, et al.
Bio-protocol, 8(4) (2018)
Chemical reaction monitoring via the light focusing in optofluidic waveguides.
Zhao HT, et al.
Sensors and Actuators B, Chemical, 280(4), 16-23 (2019)
Multi-layered, membrane-integrated microfluidics based on replica molding of a thiol-ene epoxy thermoset for organ-on-a-chip applications.
Sticker D, et al.
Lab on a chip, 15(24), 4542-4554 (2015)

Articles

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Protocols

This procedure may be used for all Catalase products.

This procedure may be used for all Ficin products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service