Skip to Content
Merck
All Photos(1)

Documents

21879

Sigma-Aldrich

5(6)-Carboxyfluorescein diacetate

BioReagent, suitable for fluorescence, ≥90.0% (HPLC)

Synonym(s):

5(6)-Carboxy-di-O-acetylfluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H16O9
CAS Number:
Molecular Weight:
460.39
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥90.0% (HPLC)

form

powder

solubility

DMSO: soluble

fluorescence

λex 492 nm; λem 517 nm in 0.1 M Tris pH 8.0 (esterase)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc2c(Oc3cc(OC(C)=O)ccc3C24OC(=O)c5ccc(cc45)C(O)=O)c1.CC(=O)Oc6ccc7c(Oc8cc(OC(C)=O)ccc8C79OC(=O)c%10cccc(C(O)=O)c9%10)c6

InChI

1S/2C25H16O9/c1-12(26)31-15-4-7-18-21(10-15)33-22-11-16(32-13(2)27)5-8-19(22)25(18)20-9-14(23(28)29)3-6-17(20)24(30)34-25;1-12(26)31-14-6-8-18-20(10-14)33-21-11-15(32-13(2)27)7-9-19(21)25(18)22-16(23(28)29)4-3-5-17(22)24(30)34-25/h2*3-11H,1-2H3,(H,28,29)

InChI key

SWRGCNMDGRMQGB-UHFFFAOYSA-N

General description

5(6)-Carboxyfluorescein diacetate or CDFDA is a fluorescent oxidant sensing probe. It is an intracellular probe added to the cells as a pro-substrate, which upon hydrolysis by the esterase enzyme produces the fluorescent product called CDF.

Application

5(6)-Carboxyfluorescein diacetate (CFDA) is a dye which is been extensively utilized to label cultured neurons with the essential requirements, in a specific manner.

5(6)- Carboxyfluorescein diacetate (CFDA) is a viability probe that assesses enzymatic activity, which is necessary to activate its fluorescence, and cell-membrane integrity, which is necessary for the retention of their fluorescent product within the cell.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsui-Chun Hsu et al.
PloS one, 13(4), e0194866-e0194866 (2018-04-12)
Zearalenone (ZEN) is a mycotoxin produced by Fusarium species, which is one of the main animal feed contaminants causing reproductive disorders in livestock. The aim of this study was to evaluate the probiotic characteristics and ZEN removal ability of a
Rodrigo Anibal Mateos-Nava et al.
Drug and chemical toxicology, 40(4), 457-462 (2016-12-13)
Vanadium is a widely distributed metal in the Earth's surface and is released into the environment by either natural or anthropogenic causes. Vanadium (III) oxide (V
Ping Li et al.
The New phytologist, 215(2), 699-710 (2017-04-07)
The interactions of vector-virus-plant have important ecological and evolutionary implications. While the tripartite interactions have received some attention, little is known about whether vector infestation affects subsequent viral transmission and infection. Working with the whitefly Bemisia tabaci, begomovirus and tobacco/tomato
Zhuo Chen et al.
Journal of controlled release : official journal of the Controlled Release Society, 320, 469-483 (2020-01-29)
Astrocytes are key stromal components in glioblastoma (GBM) and have complex interactions with the GBM cells (GBC) promoting the survival, progression and therapy resistance of GBM. In this study, we first demonstrated the existence of a reciprocal activation loop mediated
Ting Yang et al.
Immunobiology, 218(4), 548-553 (2012-08-14)
Donor lymphocyte transfusion (DLT) may induce the graft-versus-leukemia (GVL) effect for patients with AML relapsed after transplant. However, AML is a highly diverse disease and the limited overall efficacy of DLT in clinical practice emphasizes the importance of identifying a

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service