Skip to Content
Merck
All Photos(1)

Key Documents

D8189

Sigma-Aldrich

Bis(1,3-dibutylbarbituric acid) trimethine oxonol

≥95% purity (HPLC), powder or solid

Synonym(s):

DiBAC4(3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40N4O6
CAS Number:
Molecular Weight:
516.63
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Bis(1,3-dibutylbarbituric acid) trimethine oxonol, ≥95% (HPLC)

Quality Level

Assay

≥95% (HPLC)

form

powder or solid

technique(s)

titration: suitable

color

orange to dark red

solubility

DMF: soluble
DMSO: soluble

ε (extinction coefficient)

≥90000 at 490-496 nm in methanol

fluorescence

λex 513-519 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

CCCCN(C(N(CCCC)C1=O)=O)C(C1=C/C=C/C2C(N(CCCC)C(N(CCCC)C2=O)=O)=O)=O

InChI

1S/C27H40N4O6/c1-5-9-16-28-22(32)20(23(33)29(26(28)36)17-10-6-2)14-13-15-21-24(34)30(18-11-7-3)27(37)31(25(21)35)19-12-8-4/h13-15,20H,5-12,16-19H2,1-4H3/b14-13+

InChI key

ATJCYSYHWGQAET-BUHFOSPRSA-N

Application

Application in high throughput screening.
Bis(1,3-dibutylbarbituric acid) trimethine oxonol has been used for monitoring changes in the plasma membrane potential.[1][2] It also has applications in high throughput screening.

Biochem/physiol Actions

Bis(1,3-dibutylbarbituric acid) trimethine oxonolis a translational membrane potential dye that is redistributed in the cell as membrane potential changes.
Translational membrane potential dye that is redistributed in the cell as membrane potential changes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Bernabò et al.
Scientific reports, 9(1), 8155-8155 (2019-06-01)
Graphene Oxide (GO) is a widely used biomaterial with an amazing variety of applications in biology and medicine. Recently, we reported the ability of GO to improve the in vitro fertilization (IVF) outcomes in swine, a validated animal model with
Johannes Kassmannhuber et al.
Bioengineered, 8(5), 488-500 (2017-01-26)
In a concept study the ability to induce heterogeneous ice formation by Bacterial Ghosts (BGs) from Escherichia coli carrying ice nucleation protein InaZ from Pseudomonas syringae in their outer membrane was investigated by a droplet-freezing assay of ultra-pure water. As
Easa Nagamalleswari et al.
Nucleic acids research, 45(14), 8423-8434 (2017-09-01)
Programmed cell death (PCD) under certain conditions is one of the features of bacterial altruism. Given the bacterial diversity and varied life style, different PCD mechanisms must be operational that remain largely unexplored. We describe restriction endonuclease (REase) mediated cell
T Bräuner et al.
Biochimica et biophysica acta, 771(2), 208-216 (1984-04-11)
The usefulness of a new voltage-sensitive fluorescent dye, the membrane permeant negatively charged oxonol dye diBA-C4-(3)-, was evaluated by measuring the membrane potentials of BICR/M1R-k and L cells with glass microelectrodes and simultaneously recording the fluorescence of the stained cells.
B Lee et al.
Journal of applied microbiology, 127(3), 701-712 (2019-06-20)
Metal nanoparticles are promising materials for the management of infectious diseases as known to have various antimicrobial activities in pathogenic micro-organisms. Among them, gold nanoparticles (AuNPs) are used in a wide range of fields such as photodynamic therapy, molecular diagnostics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service