Skip to Content
Merck
All Photos(1)

Key Documents

14562

Sigma-Aldrich

2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein tetrakis(acetoxymethyl) ester

BioReagent, for fluorescence

Synonym(s):

BCECF-AM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H36O19
Molecular Weight:
808.69
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.32

grade

for fluorescence

product line

BioReagent

form

solid

solubility

DMF: soluble
DMSO: soluble
acetonitrile: soluble

fluorescence

λex 482 nm; λem 528 nm in 0.1 M Tris pH 8.0 (esterase)

suitability

suitable for fluorescence

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

An uncharged, nonfluorescent molecule that is useful for noninvasive bulk loading of cell suspension because it is capable of permeating the cell membrane. Cellular nonspecific esterases cleave its lipophilic blocking groups, resulting in a charged, fluorescent form of the molecule. Compared to the parent compound, this charged derivative leaks out of the cell at a much slower rate. Utilized for pH measurements in perfused tissues, intercellular spaces, mammalian cells, plant cells, bacteria, and yeast. Also, employed to investigate in assays for cellular functional properties such as adhesion, multi-drug resistance, viability and cytotoxicity, apoptosis and chemotaxis.
Cell-permeable ester of BCECF that, on hydrolysis by cytosolic esterases, yields the intracellularly trapped pH-indicator BCECF. Allows simultaneous recording of cell volume changes and intracellular pH in single osteosarcoma cells.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Sellers et al.
Journal of immunological methods, 172(2), 255-264 (1994-06-24)
A cytotoxicity assay has been developed based on the measurement of the proliferative activity of surviving cells as quantified by a cell-incorporated fluorescent dye, 2',7'-bis-(2-carboxyethyl)-5(6)-carboxyfluorescein (BCECF). The BCECF proliferative assay is fast (the results are obtained within 3-4 days depending
S Goelz et al.
The Journal of biological chemistry, 269(2), 1033-1040 (1994-01-14)
The mammalian cDNA encoding alpha (1,3)-fucosyltransferase (alpha (1,3)Fuc-T) termed ELAM-1 ligand fucosyltransferase (ELFT) or Fuc-TIV was previously cloned by three groups who reported different results from transfection studies Goelz et al. (Goelz, S. E., Hession, C., Goff, D., Griffiths, B.
A M Paradiso et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(23), 7436-7440 (1984-12-01)
We have used the pH-sensitive, fluorescent, cytoplasmic-trapped dye 2',7'-bis(carboxyethyl)-5(6)-carboxyfluorescein (BCECF) to identify Na+-H+ exchange in gastric glands isolated from rabbit stomachs by high-pressure perfusion and collagenase digestion. The fluorescence of BCECF-loaded glands was calibrated in terms of cytosolic pH (pHc)
G R Martin et al.
Cancer research, 54(21), 5670-5674 (1994-11-01)
The tumor interstitial pH and its modification play a significant role in cancer treatment. Current in vivo pH measurement techniques are invasive and/or provide poor spatial resolution. Therefore, there are no data on perivascular interstitial pH gradients in normal or
H W van Veen et al.
The Journal of biological chemistry, 269(47), 29509-29514 (1994-11-25)
The strictly aerobic, polyphosphate-accumulating Acinetobacter johnsonii strain 210A degrades its polyphosphate when oxidative phosphorylation is impaired. The endproducts of this degradation, divalent metal ions and inorganic phosphate, are excreted as a neutral metal-phosphate (MeHPO4) chelate via the electrogenic MeHPO4/H+ symport

Articles

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service