Skip to Content
Merck
All Photos(1)

Key Documents

21882

Sigma-Aldrich

5(6)-Carboxy-2′,7′-dichlorofluorescein

BioReagent, suitable for fluorescence, ≥95% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H10Cl2O7
CAS Number:
Molecular Weight:
445.21
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥95% (TLC)

form

solid

pKa 

5.1

mp

≥250 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
H2O: soluble (pH ≥ 5)

fluorescence

λex 504 nm; λem 529 nm in 0.1 M Tris pH 8.0

suitability

suitable for fluorescence

SMILES string

OC(=O)c1ccc(c(c1)C(O)=O)C2=C3C=C(Cl)C(=O)C=C3Oc4cc(O)c(Cl)cc24.OC(=O)c5ccc(C(O)=O)c(c5)C6=C7C=C(Cl)C(=O)C=C7Oc8cc(O)c(Cl)cc68

InChI

1S/2C21H10Cl2O7/c22-13-4-11-17(6-15(13)24)30-18-7-16(25)14(23)5-12(18)19(11)10-3-8(20(26)27)1-2-9(10)21(28)29;22-13-4-11-17(6-15(13)24)30-18-7-16(25)14(23)5-12(18)19(11)9-2-1-8(20(26)27)3-10(9)21(28)29/h2*1-7,24H,(H,26,27)(H,28,29)

InChI key

WHFHRELKGQJJBQ-UHFFFAOYSA-N

Application

5(6)-Carboxy-2′,7′-dichlorofluorescein may be used as a specific substrate/probe to study the identity, activity and specificity of multidrug resistance protein 2 (MRP2) molecules.
suitable as pH-indicator

Other Notes

Fluorescent intracellular pH indicator in the pH 4-5 range

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amélie Moreau et al.
Fundamental & clinical pharmacology, 25(6), 743-752 (2011-01-08)
Macrophages represent major cellular targets of various drugs, especially antibiotics and anti-viral drugs. Factors that may govern intracellular accumulation of drugs in these cells, especially those related to activity of drug transporters, are consequently likely important to consider. The present
Kati-Sisko Vellonen et al.
Journal of pharmaceutical sciences, 99(2), 1087-1098 (2009-07-23)
ATP-binding cassette (ABC) transporters are able to efflux their substrate drugs from the cells. We compared expression of efflux proteins in normal human corneal epithelial tissue, primary human corneal epithelial cells (HCEpiC), and corneal epithelial cell culture model (HCE model)
Christian Lechner et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 75(2), 284-290 (2010-03-24)
To establish a fluorescence-based assay for drug interactions with the ABC-export-protein MRP2 (ABCC2). Apical membrane vesicles were isolated by differential centrifugation from polarized MDCKII cells and MDCKII cells transfected with human MRP2. Vesicle fractions were characterized by electron microscopy, determination
Jannik Vindeløv et al.
Yeast (Chichester, England), 19(5), 429-439 (2002-03-29)
The effect of hyperosmotic shock on cell volume, vacuole volume, and intracellular pH (pH(i)) of individual cells of Saccharomyces cerevisiae and Zygosaccharomyces mellis was investigated. After transfer from a high water activity (a(w)) medium to low a(w) media, the growth
L Payen et al.
Biochemical pharmacology, 60(12), 1967-1975 (2000-12-08)
Organic anion secretion by human hepatocytes was characterized using primary liver parenchymal cell cultures and the anionic fluorescent dye carboxy-2',7'-dichlorofluorescein (CF). Probenecid, a well-known common blocker of the membrane transport process for anions, was shown to increase CF accumulation in

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service