Skip to Content
Merck
All Photos(3)

Key Documents

537993

Sigma-Aldrich

1-Butanol

99.9%

Synonym(s):

n-Butanol, Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
969148
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
bp:
116-118 °C (lit.)
vapor pressure:
5.5 mmHg
Pricing and availability is not currently available.

vapor density

2.55 (vs air)

Quality Level

vapor pressure

5.5 mmHg

Assay

99.9%

form

liquid

autoignition temp.

649 °F

expl. lim.

11.2 %

impurities

≤0.005% Acidity (as Acetic acid)
≤0.020% n-Butyraldehyde
≤0.05 mg/mL Nonvolatile matter
≤0.050% Carbonyl (as C=O)
≤0.1% water (Karl Fischer)
≤0.100% Di-n-butyl ether
≤0.100% Isobutyl alcohol

color

APHA: ≤10

refractive index

n20/D 1.399 (lit.)

pH

7 (20 °C, 70 g/L)

bp

116-118 °C (lit.)

mp

−90 °C (lit.)

solubility

water: soluble

density

0.81 g/mL at 25 °C (lit.)

cation traces

Fe: ≤0.3 ppm

SMILES string

CCCCO

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butanol is an industrial solvent that can be obtained either from petroleum feedstock or by biomass fermentation. Some of its characteristic properties include high energy content, low vapor pressure and a better capability to combine with gasoline.[1] Synthesis of di-n-butyl ether (DBE) in a fixed-bed flow reactor via dehydration of 1-butanol in the presence of γ-alumina, zeolites, and Amberlyst ion-exchange resins has been investigated.[2] Its thermophysical properties have been calculated.[3] Formation of fatty acid butyl esters (FABE) via transesterification of rapeseed and jatropha oil with 1-butanol in the presence of acid catalyst has been studied under varying conditions.[4]

Application

1-Butanol may be used for the esterification of lactic acid in the presence of lipase from Candida antarctica and hexane solvent.[5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Heat Capacity of 1-Butanol over a Wide Temperature Range.
Safarov J, et al.
Chemie Ingenieur Technik, 86(9), 1358-1358 (2014)
Salting-out of acetone, 1-butanol, and ethanol from dilute aqueous solutions.
Xie S, et al.
AIChE Journal, 61(10), 3470-3478 (2015)
Optimization of acid catalyzed transesterification of jatropha and rapeseed oil with 1-butanol.
Bynes AN, et al.
Fuel: The Science and Technology of Fuel and Energy, 137, 94-99 (2014)
Flow reactor synthesis of cetane-enhancing fuel additive from 1-butanol.
Samoilov VO, et al.
Fuel Processing Technology, 140, 312-323 (2015)
Lipase catalyzed esterification of lactic acid.
From M, et al.
Biotechnology Letters, 19(4), 315-318 (1997)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Bonjour, Quelle est la date limite de consommation ? Il y a t'il un péremption pour ce produit ? Qu'elle est sa durée de vie ? Merci d'avance pour votre retour

    1 answer
    1. This product does not contain an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service