Skip to Content
Merck
All Photos(2)

Key Documents

131377

Sigma-Aldrich

1,10-Phenanthroline

≥99%

Synonym(s):

o-phenanthroline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2
CAS Number:
Molecular Weight:
180.21
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39210124
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥99%

form

powder

mp

114-117 °C (lit.)

SMILES string

c1cnc2c(c1)ccc3cccnc23

InChI

1S/C12H8N2/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1/h1-8H

InChI key

DGEZNRSVGBDHLK-UHFFFAOYSA-N

Gene Information

human ... FNTA(2339)

Application

1,10-Phenanthroline can be used as:
  • A cathode buffer layer to improve the efficiency of organic solar cells.[1]
  • A conventional chelator to study its efficacy in Fenton′s reaction-luminol chemiluminescence system.[2]
  • A ligand in mild, copper (II)-catalyzed cross-coupling of organoboronic acids and sulfinate salts, leading to aryl- and alkenylsulfones.[3]
  • A versatile ligand employed in the spectrophotometric determination of metals[4] and photocatalytic reduction of carbon dioxide.[5]
  • A building block for metallomacrocycles.[6]

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neville S Ng et al.
Dalton transactions (Cambridge, England : 2003), 42(9), 3196-3209 (2012-12-13)
Copper(II) (1(Cu)-21(Cu)) and previously established experimental anticancer platinum(II) metallointercalator complexes (1(Pt)-16(Pt)) have been prepared and investigated for their antimicrobial properties. These complexes are of the general structure [M(I(L))(A(L))](2+) where I(L) represents functionalised 1,10-phenanthrolines (1(IL)-10(IL)), and A(L) represents 1,2-diaminoethane, 1S,2S- or
Catalysis by Metal Complexes, 14, 217-217 (1993)
Aswan Sci. Technol. Bull., 13, 3-3 (1992)
Mitsuhiro Wada et al.
Luminescence : the journal of biological and chemical luminescence, 29(7), 955-958 (2014-01-10)
In vitro screening of a Fe(2+) -chelating effect using a Fenton's reaction-luminol chemiluminescence (CL) system is described. The luminescence between the reactive oxygen species generated by the Fenton's reaction and luminol was decreased on capturing Fe(2+) using a chelator. The
Tetrahedron, 63, 7667-7667 (2007)

Questions

1–7 of 7 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Do you have GMP pharmaceutical grade 1,10-Phenanthroline?

    1 answer
    1. Unfortunately, there are no GMP or pharmaceutical grade products available at this time.

      Helpful?

  4. What is the difference between 1,10-Phenanthroline Products 131377 and 320056?

    1 answer
    1. Product 320056 is the monohydrate form and 131377 has no specification on water content. Both can make a copper complex, since they are the same ligand.

      Helpful?

  5. What is the solubility of Product 131377, 1,10-Phenanthroline?

    1 answer
    1. 1,10-Phenanthroline is soluble in methanol at 500 micrograms per mL, or in water at 3 milligrams per mL. It is also soluble in benzene and acetone.

      Helpful?

  6. Can Product 131377, 1,10-Phenanthroline, be used for cell culture?

    1 answer
    1. Sigma-Aldrich does not have any references for the use of 1,10-phenanthroline in cell culture.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service