Skip to Content
Merck
All Photos(3)

Documents

360465

Sigma-Aldrich

1-Butanol

ACS reagent, ≥99.4%

Synonym(s):

n-Butanol, Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
969148
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

2.55 (vs air)

vapor pressure

5.5 mmHg

Assay

≥99.4%

form

liquid

autoignition temp.

649 °F

expl. lim.

11.2 %

impurities

≤0.0008 meq/g Titr. acid
≤0.01% butyraldehyde
≤0.1% water
≤0.2% butyl ether

evapn. residue

≤0.005%

color

APHA: ≤10

refractive index

n20/D 1.399 (lit.)

pH

7 (20 °C, 70 g/L)

bp

116-118 °C (lit.)

mp

−90 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

SMILES string

CCCCO

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butanol is the primary alcohol that can be used as a carrier liquid for the extraction of synthetic resins, natural resins, paints, essential oils, gums, coatings, dyes, camphor, and alkaloids. It can be used as an extracting solvent for hormones, vitamins, and antibiotics. In the production of paint removers and industrial cleaners 1-butanol is being used. It is also used in the manufacture of rubber products, plastics, coating solvents, and dibutyl phthalate.

Application

1-Butanol can be used:
  • As a solvent along with the water in the delignification of sugar cane bagasse.
  • As a both reagent and reaction medium in the synthesis of zinc oxide nanoparticles from zinc acetylacetonate hydrate as precursor.
  • As an additive in the diesel-biodiesel-butanol tertiary blend which leads to fuel sustainability.
  • As a reactant in the enzyme catalyzed esterification reaction of oleic acid to butyl oleate.

greener alternative product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E Gavini et al.
International journal of pharmaceutics, 307(1), 9-15 (2005-11-01)
The nasal route is used both for local therapies and, more recently, for the systemic administration of drugs, as well as for the delivery of peptides and vaccines. In this study the nasal administration of Carbamazepine (CBZ) has been studied
The kinetics of the triethylamine-catalyzed reaction of diisocyanates with 1-butanol in toluene.
Burkus J and Eckert CF.
Journal of the American Chemical Society, 80(22), 5948-5950 (1958)
Acid-base properties of silica-aluminas: use of 1-butanol dehydration as a test reaction.
Berteau P, et al.
Applied Catalysis, 70(1), 307-323 (1991)
Prasenjit Seal et al.
The journal of physical chemistry. A, 117(2), 275-282 (2012-12-19)
In the present work, we study the H atom abstraction reactions by hydroxyl radical at all five sites of 1-butanol. Multistructural variational transition state theory (MS-VTST) was employed to estimate the five thermal rate constants. MS-VTST utilizes a multifaceted dividing
C R Shen et al.
Metabolic engineering, 10(6), 312-320 (2008-09-09)
Production of higher alcohols via the keto-acid intermediates found in microorganism's native amino-acid pathways has recently shown promising results. In this work, an Escherichia coli strain that produces 1-butanol and 1-propanol from glucose was constructed. The strain first converts glucose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service