Skip to Content
Merck
All Photos(3)

Key Documents

398268

Sigma-Aldrich

1-Pentanol

ACS reagent, ≥99%

Synonym(s):

n-Amyl alcohol, Pentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730975
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
136-138 °C (lit.)
vapor pressure:
1 mmHg ( 13.6 °C)
1.5 mmHg ( 20 °C)

grade

ACS reagent

Quality Level

vapor density

3 (vs air)

vapor pressure

1 mmHg ( 13.6 °C)
1.5 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

572 °F

expl. lim.

10 %, 100 °F

dilution

(for analytical testing)

impurities

≤0.075 meq/g acids and esters
≤0.1% carbonyl (as HCHO)
≤0.5% water

evapn. residue

≤0.003%

color

APHA: ≤30

refractive index

n20/D 1.409 (lit.)

bp

136-138 °C (lit.)

mp

−78 °C (lit.)

solubility

water: soluble 22.8 g/L at 25 °C

density

0.811 g/mL at 25 °C (lit.)

SMILES string

CCCCCO

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Pentanol is a monohydroxy alcohol. It is a colorless liquid with a characteristic odor. Its combustion properties have been studied as it shows promising features to be an alternative to gasoline and diesel fuels. The conversion of 1-pentanol to di-n-pentyl ether (DNPE) in liquid phase at 423K in a batch reactor using ion exchange resins as catalysts.

Application

1-Pentanol was used in the synthesis of 2-methyltetrahydrofuran by oxidation using ceric ammonium nitrate (CAN).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation of organic compounds with cerium (IV). IX. Formation of 2-methyltetrahydrofuran by oxidation of 1-pentanol.
Trahanovsky WS, et al.
Tetrahedron Letters, 10(30), 2501-2504 (1969)
Liquid-phase dehydration of 1-octanol, 1-hexanol and 1-pentanol to linear symmetrical ethers over ion exchange resins.
Casas C, et al.
Applied Catalysis A: General, 396(1), 129-139 (2011)
Solvothermal synthesis of submillimeter ultralong hydroxyapatite nanowires using a calcium oleate precursor in a series of monohydroxy alcohols.
Jiang YY, et al.
Ceramics International, 41(4), 6098-6102 (2015)
Experimental and detailed kinetic modeling study of 1-pentanol oxidation in a JSR and combustion in a bomb.
Togbe C, et al.
Proceedings of the Combustion Institute, 33(1), 367-374 (2011)
Anthony F Cann et al.
Applied microbiology and biotechnology, 85(4), 893-899 (2009-10-28)
Pentanol isomers such as 2-methyl-1-butanol and 3-methyl-1-butanol are a useful class of chemicals with a potential application as biofuels. They are found as natural by-products of microbial fermentations from amino acid substrates. However, the production titer and yield of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service