Skip to Content
Merck
All Photos(1)

Documents

B0600000

Benzocaine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Aminobenzoic acid ethyl ester, Ethyl 4-aminobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Beilstein:
638434
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

benzocaine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)c1ccc(N)cc1

InChI

1S/C9H11NO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6H,2,10H2,1H3

InChI key

BLFLLBZGZJTVJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzocaine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L A Linares et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 18(3), 267-269 (1990-07-01)
Methemoglobinemia is a condition in which more than 2% of the hemoglobin of the blood has been oxidized to the ferric form, a molecule which is incapable of binding oxygen or carbon dioxide. Clinically, the patient appears lethargic, cyanotic, and
Methemoglobinemia: a complication after fiberoptic orotracheal intubation with benzocaine spray. A case report.
F D Ellis et al.
The Journal of bone and joint surgery. American volume, 77(6), 937-939 (1995-06-01)
H A Cooper
Southern medical journal, 90(9), 946-948 (1997-09-26)
Acute methemoglobinemia is a medical emergency that can rapidly become fatal. A substantial number of drugs, including topical preparations, can precipitate this condition. I report a rare example of severe, acute methemoglobinemia caused by oropharyngeal anesthesia with topical benzocaine spray
Benzocaine-induced methemoglobinemia in the pediatric population.
Jodi Lehr et al.
Journal of pediatric nursing, 27(5), 583-588 (2012-07-24)
L F Rodriguez et al.
The Annals of pharmacotherapy, 28(5), 643-649 (1994-05-01)
To report a case of benzocaine-induced inethemoglobinemia and present a review of the related literature. An 83-year-old man received benzocaine topical anesthesia 600 mg prior to intubation for resection of a thyroid adenoma. The patient became severely cyanotic after induction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service