112909
Ethyl 4-aminobenzoate
98%
Synonym(s):
Et-PABA, Ethyl p-aminobenzoate
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Linear Formula:
H2NC6H4CO2C2H5
CAS Number:
Molecular Weight:
165.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
Quality Level
Assay
98%
form
solid
mp
88-90 °C (lit.)
functional group
ester
SMILES string
CCOC(=O)c1ccc(N)cc1
InChI
1S/C9H11NO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6H,2,10H2,1H3
InChI key
BLFLLBZGZJTVJG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Vibrational spectral investigations of ethyl 4-aminobenzoate in solid phase have been reported. It affords an electroactive conducting copolymer with o-anisidine, via an electrochemical synthesis employing cyclic voltammetry.
Application
Ethyl 4-aminobenzoate may be used in the synthesis of ethyl 4-[(E)-(2-hydroxy-4-methoxyphenyl)methyleneamino] benzoate (Schiff base), via reaction with 2-hydroxy-4-methoxybenzaldehyde.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
233.6 °F
Flash Point(C)
112 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Electrochemical Synthesis and Characterization of Nano Poly (o-anisidine-co-ethyl 4-aminobenzoate).
Sasikumar R and Manisankar P.
Advanced Materials Research, 678, 239-243 (2013)
Elena Pahonțu et al.
Molecules (Basel, Switzerland), 20(4), 5771-5792 (2015-04-08)
A novel Schiff base, ethyl 4-[(E)-(2-hydroxy-4-methoxyphenyl)methylene-amino]benzoate (HL), was prepared and structurally characterized on the basis of elemental analyses, (1)H NMR, (13)C NMR, UV-Vis and IR spectral data. Six new copper(II) complexes, [Cu(L)(NO3)(H2O)2] (1), [Cu(L)2] (2), [Cu(L)(OAc)] (3), [Cu2 (L)2Cl2(H2O)4] (4)
S Muthu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 112, 169-181 (2013-05-15)
The FT-IR and FT-Raman spectra of ethyl 4-aminobenzoate (EAB) in the solid phase were recorded. The fundamental vibrational wavenumbers, intensities of vibrational bands and the optimized geometrical parameters of the compound were evaluated using DFT (B3LYP) method with 6-311++G(d,p) basis
Qing Zhu et al.
Molecular pharmaceutics, 9(3), 546-553 (2012-02-04)
The crystallization kinetics of various active pharmaceutical ingredient/polyethylene glycol (API/PEG) solid dispersions has been investigated using wide-angle X-ray diffraction (XRD) and Raman spectroscopy. APIs with different physicochemical properties and crystallization tendency were employed to form solid dispersions with PEG. The
Sherry I Bame et al.
American journal of preventive medicine, 43(6 Suppl 5), S435-S442 (2012-11-28)
Hurricanes Katrina and Rita struck the Gulf Coast forcing unprecedented mass evacuation and devastation. Texas 2-1-1 is a disaster communication hub between callers with unmet needs and community services at disaster sites and evacuation destinations. To describe the location and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service