Skip to Content
Merck
All Photos(1)

Key Documents

1054000

USP

Benzocaine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Aminobenzoic acid ethyl ester, Ethyl 4-aminobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Beilstein:
638434
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

benzocaine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)c1ccc(N)cc1

InChI

1S/C9H11NO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6H,2,10H2,1H3

InChI key

BLFLLBZGZJTVJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzocaine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Benzocaine Lozenges
  • Benzocaine Gel
  • Benzocaine Cream
  • Benzocaine Ointment
  • Benzocaine Topical Aerosol®
  • Benzocaine Otic Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Legal Information

Aerosol is a registered trademark of Cytec Technology Corp.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephan Warnke et al.
Journal of the American Chemical Society, 137(12), 4236-4242 (2015-03-12)
The immediate environment of a molecule can have a profound influence on its properties. Benzocaine, the ethyl ester of para-aminobenzoic acid that finds an application as a local anesthetic, is found to adopt in its protonated form at least two
Paul M Forlano et al.
PloS one, 10(4), e0121914-e0121914 (2015-04-08)
In seasonal breeding vertebrates, hormone regulation of catecholamines, which include dopamine and noradrenaline, may function, in part, to modulate behavioral responses to conspecific vocalizations. However, natural seasonal changes in catecholamine innervation of auditory nuclei is largely unexplored, especially in the
Frédéric G Brunet et al.
BMC evolutionary biology, 15, 22-22 (2015-04-17)
The A Disintegrin-like and Metalloproteinase domain with Thrombospondin-1 motifs (ADAMTS) enzymes comprise 19 mammalian zinc-dependent metalloproteinases (metzincins) with homologues in a wide range of invertebrates. ADAMTS enzymes have a broad range of functions in development and diseases due to their
Sarmed S Sami et al.
The American journal of gastroenterology, 110(1), 148-158 (2014-12-10)
The objective of this study was to compare participation rates and clinical effectiveness of sedated esophagogastroduodenoscopy (sEGD) and unsedated transnasal endoscopy (uTNE) for esophageal assessment and Barrett's esophagus (BE) screening in a population-based cohort. This was a prospective, randomized, controlled
Tamsyn M Uren Webster et al.
BMC genomics, 16, 32-32 (2015-02-01)
Glyphosate, the active ingredient in Roundup formulations, is the most widely used herbicide worldwide, and as a result contaminates surface waters and has been detected in food residues, drinking water and human urine, raising concerns for potential environmental and human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service