Skip to Content
Merck
All Photos(3)

Key Documents

239313

Sigma-Aldrich

Sodium sulfate

Sodium sulfate

ACS reagent, ≥99.0%, anhydrous, granular

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
granular
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous

Assay

≥99.0%

form

granular

impurities

≤0.01% insolubles
≤5 ppm N compounds

loss

≤0.5% loss on ignition

pH

5.2-9.2 (25 °C, 5%)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%

cation traces

Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES)

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium sulfate (anhydrous) can be used as a catalyst in the synthesis of 1,3-diaryl-2-propen-1-one oxime by condensation of 1,3-diaryl-2-propen-1-one with hydroxylamine hydrochloride in ethanol.[1]
NaSO4 can also be used as a drying agent in organic synthesis for the removal of traces of water from organic extracts. It is used for drying almost all compounds including fatty acids, aldehydes, ketones, and alkyl and aryl halogenides.[2][3]

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 3-and 5-Alkyl-6-alkyl (aryl) tetrahydropyran-2, 4-diones by the Condensation of ?-Oxo Acid Esters with Aldehydes and Ketones
Lokot, IP, et al.
Chemistry of Heterocyclic Compounds, 37(6), 707-714 (2001)
Chromatographic characterization of the crambe (Crambe abyssinica Hochst) oil and modeling of some parameters for its conversion in biodiesel
Silveira EV, et al.
Industrial Crops and Products, 97, 545-551 (2017)
Improved Synthesis of 1, 3-Diaryl-2-propen-1-one Oxime in the Presence of Anhydrous Sodium Sulfate
Xu Xiaoya, et al.
Chin. J. Chem., 29(12), 2781-2784 (2011)
Eagleson M.
Concise Encyclopedia Chemistry, 405-405 (1994)
The use of sodium sulfate as the globulin precipitant in the determination of proteins in blood.
Howe PE.
The Journal of Biological Chemistry, 49(1), 93-107 (1921)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service