Skip to Content
Merck
All Photos(1)

Key Documents

A6848

Supelco

Aztreonam

analytical standard

Synonym(s):

[2S-[2α,3β(Z)]]-2-[[[1-(2-Amino-4-thiazolyl)-2-[(2-methyl-4-oxo-1-sulfo-3-azetidinyl)amino]-2-oxoethylidene]amino]oxy]-2-methylpropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17N5O8S2
CAS Number:
Molecular Weight:
435.43
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

form

powder or crystals

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

SMILES string

C[C@H]1[C@H](NC(=O)\C(=N/OC(C)(C)C(O)=O)c2csc(N)n2)C(=O)N1S(O)(=O)=O

InChI

1S/C13H17N5O8S2/c1-5-7(10(20)18(5)28(23,24)25)16-9(19)8(6-4-27-12(14)15-6)17-26-13(2,3)11(21)22/h4-5,7H,1-3H3,(H2,14,15)(H,16,19)(H,21,22)(H,23,24,25)/b17-8-/t5-,7-/m0/s1

InChI key

WZPBZJONDBGPKJ-VEHQQRBSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aztreonam belongs to the group of monocyclic β-lactam antibiotics, isolated from Chromobacterium violaceum. It is used against gram-negative bacterial infections, such as infections of the bladder, kidneys, bone, and urinary tract. It acts by binding to the penicillin-binding protein-3 (PBP3) and inhibiting the synthesis of bacterial cell walls causing cell lysis and death.

Application

The analytical standard can be used as follows:
  • Determination of aztreonam in two simulated lung fluid solutions― artificial lysosomal fluid and gamble solution by a UV spectroscopic method
  • Multi-residue analysis of eight β-lactams by ultra-high performance liquid chromatography (UHPLC) combined with a photo-diode array (PDA) detector in human plasma & serum samples
  • Simultaneous quantitative analysis of nine β-lactams by UHPLC along with tandem mass spectrometry (MS/MS) in human plasma samples

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Antimicrobial Drugs (2000)
Neonatal and Pediatric Pharmacology: Therapeutic Principles in Practice (2010)
W C Hellinger et al.
Mayo Clinic proceedings, 74(4), 420-434 (1999-04-30)
Imipenem and meropenem, members of the carbapenem class of beta-lactam antibiotics, are among the most broadly active antibiotics available for systemic use in humans. They are active against streptococci, methicillin-sensitive staphylococci, Neisseria, Haemophilus, anaerobes, and the common aerobic gram-negative nosocomial
Greg L Plosker
Drugs, 70(14), 1843-1855 (2010-09-15)
Aztreonam is a monobactam antibacterial with bactericidal activity against a wide range of aerobic Gram-negative bacteria, including Pseudomonas aeruginosa. Aztreonam lysine for inhalation solution (AZLI) does not contain the potentially inflammatory component arginine, which is used as a buffer in
Leigh R Wexler et al.
Current biology : CB, 30(14), 2695-2706 (2020-06-13)
Dynamic integration of internal and external cues is essential for flexible, adaptive behavior. In C. elegans, biological sex and feeding state regulate expression of the food-associated chemoreceptor odr-10, contributing to plasticity in food detection and the decision between feeding and exploration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service