Skip to Content
Merck
All Photos(1)

Key Documents

1046205

USP

Aztreonam

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

[2S-[2α,3β(Z)]]-2-[[[1-(2-Amino-4-thiazolyl)-2-[(2-methyl-4-oxo-1-sulfo-3-azetidinyl)amino]-2-oxoethylidene]amino]oxy]-2-methylpropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17N5O8S2
CAS Number:
Molecular Weight:
435.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

aztreonam

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

C[C@H]1[C@H](NC(=O)\C(=N/OC(C)(C)C(O)=O)c2csc(N)n2)C(=O)N1S(O)(=O)=O

InChI

1S/C13H17N5O8S2/c1-5-7(10(20)18(5)28(23,24)25)16-9(19)8(6-4-27-12(14)15-6)17-26-13(2,3)11(21)22/h4-5,7H,1-3H3,(H2,14,15)(H,16,19)(H,21,22)(H,23,24,25)/b17-8-/t5-,7-/m0/s1

InChI key

WZPBZJONDBGPKJ-VEHQQRBSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Aztreonam USP reference standard is meant to use for specified quality tests and assays.

Also used to prepare standard and system suitability solutions for assay according to the given below monographs of United States Pharmacopeia (USP):
  • Aztreonam
  • Aztreonam for Injection
  • Aztreonam Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aztreonam for Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(3), 463-463 (2018)
Aztreonam Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(3), 462-462 (2018)
Michael N Dudley et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 56(9), 1301-1309 (2013-01-22)
Widespread resistance in Enterobacteriaceae and Pseudomonas aeruginosa to cephalosporin and monobactam antibiotics due to extended-spectrum β-lactamases (ESBLs) has resulted in the need for reassessment of the interpretative criteria (breakpoints) established for these agents more than 2 decades ago. Following extensive
Stéphanie Nemeghaire et al.
The Journal of antimicrobial chemotherapy, 69(11), 2928-2934 (2014-07-27)
This study aimed at assessing the epidemiology and genetic diversity of methicillin-resistant Staphylococcus sciuri (MRSS) from different farm animal species. Nasal swabs were collected from 200 pigs, 100 dairy cows, 100 beef cows, 150 veal calves and 200 broilers. Colonies
Kristen Zeitler et al.
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists, 69(2), 107-115 (2012-01-05)
The pharmacology, safety, efficacy, pharmacokinetics, pharmacodynamics, current place in therapy, and potential future therapeutic uses of inhaled aztreonam are reviewed. Inhaled aztreonam, a newly formulated lysine salt of the original monobactam antibiotic, is approved for the treatment of respiratory symptoms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service