Skip to Content
Merck
All Photos(1)

Key Documents

79560

Supelco

Phosphomolybdic acid hydrate

for microscopy

Synonym(s):

Molybdophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3[P(Mo3O10)4] · xH2O
CAS Number:
Molecular Weight:
1825.25 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

for microscopy

Quality Level

form

crystals

reaction suitability

reagent type: oxidant

technique(s)

thin layer chromatography (TLC): suitable

loss

21-23% loss on drying, 110 °C

density

1.62 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤20 mg/kg
Cr: ≤5 mg/kg
Cu: ≤10 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤20 mg/kg
Mn: ≤100 mg/kg
NH4+: ≤30 mg/kg
Na: ≤50 mg/kg
Ni: ≤10 mg/kg
Pb: ≤20 mg/kg
Zn: ≤50 mg/kg

suitability

suitable for microscopy

SMILES string

O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.OP(O)(O)=O

InChI

1S/12Mo.H3O4P.H2O.36O/c;;;;;;;;;;;;1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;;;;;;;;;;;;(H3,1,2,3,4);1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

PDDXOPNEMCREGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphomolybdic acid hydrate (PAH) is yellow and crystallisable compound. It precipitates from potash, oxides of rubidium, caesium and thallium, ammonia and other nitrogenous organic alkaloids.

Application

A precursor to hybrid organic-inorganic based molecular batteries with applications as cation-insertion electrodes.
PAH was used in preparing Mo-Al2O3 catalysts in an attempt to produce ultra-low sulphur diesel.

Other Notes

Review: of the folin phenol method for the quantitative determination of proteins

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 3 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultra-deep oxidative desulfurization of diesel fuel by the Mo/Al 2 O 3-H 2 O 2 system: The effect of system parameters on catalytic activity.
Garcia-Gutierrez, Jose Luis, et al.
Applied Catalysis A: General, 334, 366-373 (2008)
Chemical News
A new precipitant for Potash, 183-183 (1868)
Review of the Folin phenol protein quantitation method of Lowry, Rosebrough, Farr and Randall.
G L Peterson
Analytical biochemistry, 100(2), 201-220 (1979-12-01)
Dieter Fink et al.
Cancers, 12(1) (2020-01-08)
The ING3 candidate tumour suppressor belongs to a family of histone modifying proteins involved in regulating cell proliferation, senescence, apoptosis, chromatin remodeling, and DNA repair. It is a stoichiometric member of the minimal NuA4 histone acetyl transferase (HAT) complex consisting
Jae-Jin Lee et al.
Current microbiology, 69(3), 286-291 (2014-04-22)
A Gram-negative, short-rod-shaped bacterial strain with gliding motility, designated as DG5A(T), was isolated from a rice field soil in South Korea. Phylogenic analysis using 16S rRNA gene sequence of the new isolate showed that strain DG5A(T) belong to the genus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service