Skip to Content
Merck
All Photos(3)

Key Documents

P2395

Sigma-Aldrich

Ponceau Xylidine

Dye content ≥60 %, Powder

Synonym(s):

Acid Red 26, Ponceau 2 R, Xylidine ponceau, Xylidine ponceau 2R

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N2Na2O7S2
CAS Number:
Molecular Weight:
480.42
Colour Index Number:
16150
Beilstein:
5709350
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Ponceau Xylidine, Dye content ≥60 %

form

powder

composition

Dye content, ≥60%

solubility

water: 10 mg/mL, clear, red

εmax

18-22 at 503-508 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Cc1ccc(\N=N\c2c(O)c(cc3cc(ccc23)S([O-])(=O)=O)S([O-])(=O)=O)c(C)c1

InChI

1S/C18H16N2O7S2.2Na/c1-10-3-6-15(11(2)7-10)19-20-17-14-5-4-13(28(22,23)24)8-12(14)9-16(18(17)21)29(25,26)27;;/h3-9,21H,1-2H3,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2/b20-19+;;

InChI key

YJVBLROMQZEFPA-LLIZZRELSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ponceau Xylidine is a red powder which is easily soluble in water. It is used as a substitute for cochineal dye.

Application

Ponceau Xylidine has been used as part of the Goldner′s trichome stain to stain thin sections. It has also been used to prepare Fuchsin-Ponceau solution to stain an amyotrophic lateral sclerosis tissue-engineered skin model (ALS-TES).

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

S.P. Sadtler
A hand-book of industrial organic chemistry (1891)
Early detection of structural abnormalities and cytoplasmic accumulation of TDP-43 in tissue-engineered skins derived from ALS patients
Bastien Pare
Acta Neuropathologica Communications, 3(5) (2015)
Kwok-Sui Leung, Wing-Hoi Cheung
A Practical Manual for Musculoskeletal Research (2008)
Shota Kurotsu et al.
The Journal of pharmacology and experimental therapeutics, 350(1), 79-88 (2014-04-29)
Idiopathic pulmonary fibrosis is thought to involve lung injury caused by reactive oxygen species (ROS), which in turn is followed by abnormal fibrosis. A transforming growth factor (TGF)-β1-induced increase in myofibroblast number plays an important role in this abnormal fibrosis.
Henriette Jülke et al.
Cell transplantation, 24(6), 1111-1125 (2013-12-18)
The purpose of this study was to evaluate the use of three different superparamagnetic iron oxide (SPIO) particles for labeling of ovine and equine bone marrow (BM)-derived multipotent stromal cells (MSCs) in vitro. MSCs were obtained from five adult sheep

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service