Skip to Content
Merck
All Photos(1)

Documents

P4869

Sigma-Aldrich

Phosphomolybdic acid solution

spray reagent, 10% in ethanol

Synonym(s):

Molybdophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3[P(Mo3O10)4]
CAS Number:
Molecular Weight:
1825.25
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: oxidant

packaging

pkg of 100 mL

concentration

10% in ethanol

storage temp.

2-8°C

SMILES string

O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.OP(O)(O)=O

InChI

1S/12Mo.H3O4P.36O/c;;;;;;;;;;;;1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;;;;;;;;;;;;(H3,1,2,3,4);;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

DHRLEVQXOMLTIM-UHFFFAOYSA-N

Application

For the detection of lipids, steroids, lactones, keto acids, hydroxy acids, unsaturated fatty acids, and phenolic compounds.

Other Notes

10% Phosphomolybdic acid in ethanol

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Ox. Liq. 3 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

55.0 °F - closed cup

Flash Point(C)

12.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rubel Chakravarty et al.
Journal of chromatography. A, 1220, 82-91 (2011-12-27)
Barium-137m availed from a (137)Cs/(137m)Ba generator is an attractive industrial radiotracer for liquid flow rate measurement owing to its short half-life (T(1/2)=2.5 min) and emission of highly energetic gamma radiation (E(γ)=661.7 keV). Ammonium molybdophosphate (AMP) impregnated alumina microspheres (AMP-Al(2)O(3)), an
Maher Mahmoud El-Hashash et al.
Acta pharmaceutica (Zagreb, Croatia), 60(3), 361-367 (2010-12-08)
In the present study, antioxidant activity of methanolic extracts of the leaves of seven Egyptian Cassia species was investigated using two methods, the phosphomolyb-date method and 1,1 diphenyl-2-picrylhydrazyl radical (DPPH) scavenging activity method.· The results revealed that C. glauca is
Dawei Fan et al.
Journal of colloid and interface science, 351(1), 151-155 (2010-08-19)
Hybrid films composed of oppositely charged Keggin-type 12-molybdophosphoric acid H(3)PMo(12)O(40).nH(2)O (PMo(12)) and water-soluble cationic meso-tetra (N-methyl-4-pyridyl) porphinetetratosylate (TMPyP) were fabricated onto silicon and ITO substrates by a layer-by-layer (LbL) self-assembly technique. The LbL films were characterized by UV-vis spectroscopy, X-ray
Rasha M El-Nashar et al.
Analytica chimica acta, 730, 99-111 (2012-05-29)
This work offers construction and comparative evaluation the performance characteristics of conventional polymer (I), carbon paste (II) and carbon nanotubes chemically modified carbon paste ion selective electrodes (III) for meclofenoxate hydrochloride are described. These electrodes depend mainly on the incorporation
Almino Afonso de O Paiva et al.
International immunopharmacology, 11(9), 1241-1250 (2011-04-21)
This study analyzes the action of sulfated polysaccharides, fucans, from algae Lobophora variegata on zymosan-induced arthritis in rats. Groups of fucans, obtained after acetone fractionation (0.3-2.0 volumes), were denominated F0.3, F0.5, F0.8, F1, F1.5, and F2. The results that F1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service